Assay Details
| Gene Symbol: LINC01185 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033980.1 | 1 - 2 | 86 | 69 | ||
| GenBank mRNA | AK122786.1 | - | 1 - 2 | 86 | 69 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | poly(A) polymerase gamma | PAPOLG | -54.7kb | Anti-sense | Chr. 2 - 60756230 - 60802086 on NCBI Build GRCh38 | |
| Downstream |
REL proto-oncogene, NF-kB subunit |
REL | C-Rel | 181.0 | Anti-sense | Chr. 2 - 60881495 - 60928171 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02001887.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01221527 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01221525 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15248183 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01221538 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21301905 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21301909 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01221474 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01221533 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21301874 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21301984 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21301911 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01221495 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21301938 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21301927 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01221549 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21301906 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21301877 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21301869 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01221506 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01221483 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01221451 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21301898 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01221472 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21301966 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01221494 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01221501 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01221559 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21301912 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21301940 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21301866 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01221478 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21301953 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21301986 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01221522 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01221505 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01221521 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21301900 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21301917 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01221500 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01221508 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01221531 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01221504 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21301960 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21301956 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15248193 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01221516 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21301988 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01221467 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01221470 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21301943 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15248184 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21301976 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21301916 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01221475 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01221512 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01221518 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21301971 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01221539 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01221479 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01221463 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01221489 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21301918 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21301965 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21301978 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01221468 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21301972 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01221481 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21301913 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01221476 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21301903 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01221507 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01221491 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01221524 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21301871 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01221466 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21301870 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21301922 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15248182 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01221499 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21301881 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01221526 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21301873 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21301904 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01221543 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01221490 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21301975 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01221509 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01221452 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15248192 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 144391 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 138371 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 39499 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 8x60K | 17556 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 956392 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 597880 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 879731 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 521338 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 363836 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 402942 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 759661 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 562162 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 903970 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 682013 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 28871 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 355530 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 466086 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 362035 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 368854 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 20904 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 98670 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 129322 | AB028975.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 129322 | AK001412.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 129322 | AL137265.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 129322 | AL833532.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 129322 | BX648682.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 129322 | NM_014956.4 | refseq_mrna_human_NM |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 99056 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3677050 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 6526 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3677050 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3677050 | - | - |
| Agilent Human Genome CGH Microarray 105A (G4412A) | A_16_P00387101 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P00387101 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P15675134 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 62226 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 166494 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 93226 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P13237160 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P35699875 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00387099 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00387095 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P15675137 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P35699926 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P15675159 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P15675134 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P15675128 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P35699907 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00387101 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00387082 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P13237615 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P35699930 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P15675156 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P15675130 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P00387082 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_18_P13237615 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P00387101 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P15675134 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P35699926 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P15675156 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02030075.hg.1 | TC02001887.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02030075.hg.1 | ENST00000452343.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02030075.hg.1 | NR_033980.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012719.hg.1 | ENST00000439412.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012719.hg.1 | ENST00000439412.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012719.hg.1 | ENST00000439412.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012719.hg.1 | ENST00000439412.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012719.hg.1 | ENST00000439412.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012719.hg.1 | ENST00000439412.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012719.hg.1 | ENST00000439412.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012719.hg.1 | ENST00000439412.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012719.hg.1 | ENST00000452343.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012719.hg.1 | ENST00000452343.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012719.hg.1 | ENST00000452343.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012719.hg.1 | ENST00000452343.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012719.hg.1 | NR_033980.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012719.hg.1 | NR_033980.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012719.hg.1 | NR_033980.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012719.hg.1 | NR_033980.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02030073.hg.1 | TC02001887.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02030073.hg.1 | ENST00000452343.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02030073.hg.1 | NR_033980.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02030074.hg.1 | TC02001887.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02030074.hg.1 | ENST00000452343.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02030074.hg.1 | NR_033980.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200192711.hg.1 | ENST00000452343.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200192711.hg.1 | NR_033980.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200192711.hg.1 | ENST00000452343.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200192711.hg.1 | NR_033980.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200192711.hg.1 | TC0200012719.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200192716.hg.1 | ENST00000439412.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200192716.hg.1 | ENST00000439412.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200192716.hg.1 | ENST00000452343.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200192716.hg.1 | NR_033980.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200192716.hg.1 | ENST00000439412.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200192716.hg.1 | ENST00000439412.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200192716.hg.1 | ENST00000452343.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200192716.hg.1 | NR_033980.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200192716.hg.1 | TC0200012719.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200192712.hg.1 | ENST00000452343.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200192712.hg.1 | NR_033980.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200192712.hg.1 | ENST00000452343.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200192712.hg.1 | NR_033980.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200192712.hg.1 | TC0200012719.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200012719.hg.1 | ENST00000439412.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012719.hg.1 | ENST00000439412.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012719.hg.1 | ENST00000439412.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012719.hg.1 | ENST00000439412.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012719.hg.1 | ENST00000439412.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012719.hg.1 | ENST00000439412.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012719.hg.1 | ENST00000439412.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012719.hg.1 | ENST00000439412.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012719.hg.1 | ENST00000452343.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012719.hg.1 | ENST00000452343.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012719.hg.1 | ENST00000452343.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012719.hg.1 | ENST00000452343.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012719.hg.1 | NR_033980.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012719.hg.1 | NR_033980.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012719.hg.1 | NR_033980.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012719.hg.1 | NR_033980.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200192715.hg.1 | ENST00000439412.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200192715.hg.1 | ENST00000439412.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200192715.hg.1 | ENST00000452343.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200192715.hg.1 | NR_033980.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200192715.hg.1 | ENST00000439412.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200192715.hg.1 | ENST00000439412.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200192715.hg.1 | ENST00000452343.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200192715.hg.1 | NR_033980.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200192715.hg.1 | TC0200012719.hg.1 | Clariom D TC |
| Human Genome U133 Plus 2.0 Array | 231630_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 231630_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A013682 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A013682 | - | - |

LINC01185
Genomic Map