Assay Details
| Gene Symbol: FLJ37505 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033987.1 | 2 - 3 | 208 | 81 | ||
| GenBank mRNA | AK094824.1 | - | 2 - 3 | 183 | 81 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC105370068 | LOC105370068 | -242.5kb | Anti-sense | Chr. 12 - 127648896 - 127655588 on NCBI Build GRCh38 | |
| Downstream | long intergenic non-protein coding RNA 507 | LINC00507 | 16.8kb | Sense | Chr. 12 - 127915410 - 127951552 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12001020.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08681633 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08681747 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08681756 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29119124 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08681652 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08681725 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08681615 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29119138 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29119151 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29119261 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08681657 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29119170 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08681731 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29119178 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29119131 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08681736 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29119188 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29119161 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08681727 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29119268 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29119274 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29119186 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08681658 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29119167 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29119123 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29119132 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08681635 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08681631 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29119182 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08681659 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08681663 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08681606 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29119145 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29119181 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08681729 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29119253 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29119152 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08681740 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08681618 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29119175 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29119159 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29119177 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29119140 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08681625 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08681735 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08681750 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29119128 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29119141 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29119166 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08681755 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08681644 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08681617 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16602681 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08681622 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08681646 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29119264 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08681593 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08681737 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29119265 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08681626 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29119129 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08681601 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08681623 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08681749 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29119180 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29119127 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08681607 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08681724 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08681757 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08681642 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08681730 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08681624 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 653408 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 280713 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 791733 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 242882 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 903982 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 135703 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3809497 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 14861 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3809497 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3809497 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P39857276 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P39857259 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P19720780 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P02724410 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P39857276 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P19532310 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P39857276 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P02724410 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR12013494.hg.1 | ENST00000536342.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12013494.hg.1 | NR_033987.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12013494.hg.1 | TC12001020.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12013497.hg.1 | NR_033987.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12013497.hg.1 | TC12001020.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009355.hg.1 | ENST00000536342.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009355.hg.1 | ENST00000536342.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009355.hg.1 | ENST00000536342.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009355.hg.1 | ENST00000536342.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009355.hg.1 | ENST00000614177.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009355.hg.1 | ENST00000614177.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009355.hg.1 | ENST00000614177.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009355.hg.1 | ENST00000614177.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009355.hg.1 | ENST00000614177.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009355.hg.1 | NR_033987.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009355.hg.1 | NR_033987.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009355.hg.1 | NR_033987.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009355.hg.1 | NR_033987.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009355.hg.1 | NR_033987.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009355.hg.1 | NR_033987.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12013498.hg.1 | NR_033987.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12013498.hg.1 | TC12001020.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1200169705.hg.1 | ENST00000536342.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200169705.hg.1 | NR_033987.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200169705.hg.1 | ENST00000536342.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200169705.hg.1 | NR_033987.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200169705.hg.1 | TC1200009355.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200169706.hg.1 | ENST00000536342.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200169706.hg.1 | ENST00000614177.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200169706.hg.1 | NR_033987.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200169706.hg.1 | ENST00000536342.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200169706.hg.1 | ENST00000614177.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200169706.hg.1 | NR_033987.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200169706.hg.1 | TC1200009355.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200169709.hg.1 | ENST00000614177.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200169709.hg.1 | NR_033987.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200169709.hg.1 | ENST00000614177.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200169709.hg.1 | NR_033987.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200169709.hg.1 | TC1200009355.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200169710.hg.1 | ENST00000614177.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200169710.hg.1 | NR_033987.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200169710.hg.1 | ENST00000614177.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200169710.hg.1 | NR_033987.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200169710.hg.1 | TC1200009355.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200169712.hg.1 | ENST00000614177.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200169712.hg.1 | NR_033987.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200169712.hg.1 | ENST00000614177.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200169712.hg.1 | NR_033987.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200169712.hg.1 | TC1200009355.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200009355.hg.1 | ENST00000536342.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009355.hg.1 | ENST00000536342.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009355.hg.1 | ENST00000536342.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009355.hg.1 | ENST00000536342.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009355.hg.1 | ENST00000614177.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009355.hg.1 | ENST00000614177.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009355.hg.1 | ENST00000614177.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009355.hg.1 | ENST00000614177.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009355.hg.1 | ENST00000614177.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009355.hg.1 | NR_033987.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009355.hg.1 | NR_033987.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009355.hg.1 | NR_033987.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009355.hg.1 | NR_033987.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009355.hg.1 | NR_033987.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009355.hg.1 | NR_033987.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200169711.hg.1 | ENST00000614177.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200169711.hg.1 | NR_033987.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200169711.hg.1 | ENST00000614177.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200169711.hg.1 | NR_033987.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200169711.hg.1 | TC1200009355.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A049193 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A049193 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 233606 | - | - |

FLJ37505
Genomic Map