Assay Details
Gene Symbol: RNF144A-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_033997.1 | 3 - 4 | 801 | 101 | ||
GenBank mRNA | AK090737.1 | - | 1 - 2 | 90 | 101 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | radical S-adenosyl methionine domain containing 2 | RSAD2 |
2510004L01Rik, cig33, cig5, vig1 |
-16.0kb | Anti-sense | Chr. 2 - 6865733 - 6898239 on NCBI Build GRCh38 |
Downstream | ring finger protein 144A | RNF144A |
RNF144, UBCE7IP4 |
1.4kb | Anti-sense | Chr. 2 - 6917392 - 7076886 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC02001544.hg.1
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0200011627.hg.1 | ENST00000418970.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011627.hg.1 | ENST00000418970.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011627.hg.1 | ENST00000418970.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011627.hg.1 | ENST00000418970.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011627.hg.1 | ENST00000418970.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011627.hg.1 | ENST00000418970.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011627.hg.1 | ENST00000418970.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011627.hg.1 | ENST00000418970.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011627.hg.1 | ENST00000418970.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011627.hg.1 | ENST00000418970.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011627.hg.1 | ENST00000418970.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011627.hg.1 | ENST00000418970.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011627.hg.1 | ENST00000418970.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011627.hg.1 | ENST00000418970.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011627.hg.1 | ENST00000418970.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011627.hg.1 | ENST00000418970.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011627.hg.1 | ENST00000418970.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011627.hg.1 | ENST00000418970.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011627.hg.1 | ENST00000418970.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011627.hg.1 | ENST00000418970.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011627.hg.1 | ENST00000418970.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011627.hg.1 | ENST00000418970.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011627.hg.1 | ENST00000418970.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011627.hg.1 | ENST00000418970.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011627.hg.1 | ENST00000418970.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011627.hg.1 | ENST00000418970.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011627.hg.1 | ENST00000424351.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011627.hg.1 | ENST00000424351.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011627.hg.1 | ENST00000424351.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011627.hg.1 | ENST00000424351.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011627.hg.1 | ENST00000424351.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011627.hg.1 | ENST00000424351.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011627.hg.1 | ENST00000424351.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011627.hg.1 | ENST00000424351.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011627.hg.1 | ENST00000424351.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011627.hg.1 | ENST00000424351.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011627.hg.1 | ENST00000424351.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011627.hg.1 | ENST00000426122.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011627.hg.1 | ENST00000426122.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011627.hg.1 | ENST00000426122.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011627.hg.1 | ENST00000426122.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011627.hg.1 | ENST00000426122.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011627.hg.1 | ENST00000426122.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011627.hg.1 | ENST00000437589.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011627.hg.1 | ENST00000437589.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011627.hg.1 | ENST00000437589.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011627.hg.1 | ENST00000437589.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011627.hg.1 | ENST00000437589.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011627.hg.1 | ENST00000439208.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011627.hg.1 | ENST00000439208.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011627.hg.1 | ENST00000439208.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011627.hg.1 | ENST00000439208.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011627.hg.1 | ENST00000439208.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011627.hg.1 | NR_033997.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011627.hg.1 | NR_033997.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011627.hg.1 | NR_033997.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011627.hg.1 | NR_033997.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011627.hg.1 | NR_033997.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011627.hg.1 | NR_033997.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011627.hg.1 | NR_033997.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011627.hg.1 | NR_033997.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011627.hg.1 | NR_033997.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011627.hg.1 | NR_033997.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011627.hg.1 | NR_033997.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011627.hg.1 | NR_033997.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02024977.hg.1 | TC02001544.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02024977.hg.1 | ENST00000418970.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02024977.hg.1 | NR_033997.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200184466.hg.1 | ENST00000418970.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200184466.hg.1 | ENST00000418970.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200184466.hg.1 | ENST00000426122.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200184466.hg.1 | NR_033997.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200184466.hg.1 | ENST00000418970.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200184466.hg.1 | ENST00000418970.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200184466.hg.1 | ENST00000426122.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200184466.hg.1 | NR_033997.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200184466.hg.1 | TC0200011627.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0200011627.hg.1 | ENST00000418970.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011627.hg.1 | ENST00000418970.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011627.hg.1 | ENST00000418970.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011627.hg.1 | ENST00000418970.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011627.hg.1 | ENST00000418970.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011627.hg.1 | ENST00000418970.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011627.hg.1 | ENST00000418970.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011627.hg.1 | ENST00000418970.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011627.hg.1 | ENST00000418970.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011627.hg.1 | ENST00000418970.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011627.hg.1 | ENST00000418970.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011627.hg.1 | ENST00000418970.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011627.hg.1 | ENST00000418970.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011627.hg.1 | ENST00000418970.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011627.hg.1 | ENST00000418970.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011627.hg.1 | ENST00000418970.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011627.hg.1 | ENST00000418970.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011627.hg.1 | ENST00000418970.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011627.hg.1 | ENST00000418970.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011627.hg.1 | ENST00000418970.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011627.hg.1 | ENST00000418970.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011627.hg.1 | ENST00000418970.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011627.hg.1 | ENST00000418970.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011627.hg.1 | ENST00000418970.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011627.hg.1 | ENST00000418970.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011627.hg.1 | ENST00000418970.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011627.hg.1 | ENST00000424351.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011627.hg.1 | ENST00000424351.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011627.hg.1 | ENST00000424351.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011627.hg.1 | ENST00000424351.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011627.hg.1 | ENST00000424351.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011627.hg.1 | ENST00000424351.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011627.hg.1 | ENST00000424351.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011627.hg.1 | ENST00000424351.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011627.hg.1 | ENST00000424351.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011627.hg.1 | ENST00000424351.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011627.hg.1 | ENST00000424351.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011627.hg.1 | ENST00000426122.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011627.hg.1 | ENST00000426122.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011627.hg.1 | ENST00000426122.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011627.hg.1 | ENST00000426122.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011627.hg.1 | ENST00000426122.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011627.hg.1 | ENST00000426122.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011627.hg.1 | ENST00000437589.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011627.hg.1 | ENST00000437589.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011627.hg.1 | ENST00000437589.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011627.hg.1 | ENST00000437589.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011627.hg.1 | ENST00000437589.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011627.hg.1 | ENST00000439208.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011627.hg.1 | ENST00000439208.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011627.hg.1 | ENST00000439208.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011627.hg.1 | ENST00000439208.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011627.hg.1 | ENST00000439208.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011627.hg.1 | NR_033997.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011627.hg.1 | NR_033997.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011627.hg.1 | NR_033997.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011627.hg.1 | NR_033997.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011627.hg.1 | NR_033997.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011627.hg.1 | NR_033997.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011627.hg.1 | NR_033997.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011627.hg.1 | NR_033997.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011627.hg.1 | NR_033997.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011627.hg.1 | NR_033997.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011627.hg.1 | NR_033997.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011627.hg.1 | NR_033997.1 | Refseq |
