Assay Details
| Gene Symbol: LOC100129148 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033999.1 | 1 - 2 | 128 | 93 | ||
| GenBank mRNA | AI811258.1 | - | 2 - 3 | 93 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | C7orf55-LUC7L2 readthrough | C7orf55-LUC7L2 | Anti-sense | Chr. 7 - 139341132 - 139423457 on NCBI Build GRCh38 | ||
| Downstream | C-type lectin domain family 2 member L | CLEC2L | 96.4kb | Anti-sense | Chr. 7 - 139523928 - 139544985 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07001913.hg.1
Set Membership: |
|

LOC100129148
Genomic Map