Assay Details
| Gene Symbol: BAALC-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_109954.1 | 1 - 2 | 201 | 167 | ||
| GenBank mRNA | AK311189.1 | - | 1 - 2 | 201 | 167 | |
| DA226722.1 | - | 1 - 2 | 202 | 168 | ||
| DA333188.1 | - | 1 - 2 | 88 | 168 | ||
| DT217610.1 | - | 1 - 2 | 88 | 167 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 3151 | MIR3151 | mir-3151 | -11.1kb | Anti-sense | Chr. 8 - 103154614 - 103154689 on NCBI Build GRCh38 |
| Downstream | frizzled class receptor 6 | FZD6 |
FZ-6, FZ6, HFZ6, NDNC10 |
126.4kb | Anti-sense | Chr. 8 - 103298433 - 103332866 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16122341 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347817 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347885 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347886 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630973 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630913 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630916 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347816 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630958 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347804 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630901 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630963 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630971 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347818 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630824 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630822 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347840 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630922 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347839 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630930 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26631009 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630941 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347819 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630903 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630966 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347791 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630957 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347881 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347823 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347793 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347870 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347834 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347703 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630813 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630949 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630942 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26631002 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630964 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630985 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347803 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347800 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630996 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347862 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347697 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347813 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347796 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630989 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630899 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347824 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630990 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347828 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347896 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630962 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630909 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347794 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630917 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630921 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630956 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630980 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347869 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347833 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347695 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630974 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347706 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630965 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630969 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347884 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347821 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347827 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347814 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630994 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347865 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630926 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630952 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347830 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347701 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630993 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26631003 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347888 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630920 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347829 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630914 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630923 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347836 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347895 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630967 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347811 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630975 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630968 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347860 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630904 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347893 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630905 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630932 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630946 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630992 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630931 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630978 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3396349 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3278205 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 60799 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 54087 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P01984377 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P18422780 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P01984391 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P01984436 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P18422812 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P18422793 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P01984391 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P18422812 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3278205 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3278205 | - | - |
| Agilent Human Genome CGH Microarray 105A (G4412A) | A_16_P01984391 | - | - |
| Agilent Human Genome CGH Microarray 105A (G4412A) | A_16_P18422812 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P01984391 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P01984436 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P18422793 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P18422812 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P18422780 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P01984377 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 136400 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 152639 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 79903 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 70500 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 30170 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P18422757 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P18422739 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P18422793 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P18422819 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P01984391 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P16608088 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P38516902 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P16608873 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P16609038 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P18422803 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P18422824 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P01984415 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P01984377 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P38516954 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P01984436 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P18422747 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P18422731 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P18422780 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P18422829 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P16607897 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P16606171 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P01984373 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P16605495 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P18422726 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P18422707 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P38516931 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P18422716 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P18422812 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P18422806 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P01984410 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P26227978 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P01984404 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800012449.hg.1 | ENST00000500902.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012449.hg.1 | ENST00000500902.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012449.hg.1 | ENST00000500902.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012449.hg.1 | ENST00000500902.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012449.hg.1 | ENST00000521102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012449.hg.1 | ENST00000521102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012449.hg.1 | ENST00000521102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012449.hg.1 | NR_109954.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012449.hg.1 | NR_109954.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012449.hg.1 | NR_109954.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012449.hg.1 | NR_109954.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800187892.hg.1 | ENST00000500902.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800187892.hg.1 | ENST00000521102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800187892.hg.1 | NR_109954.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800187892.hg.1 | ENST00000500902.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800187892.hg.1 | ENST00000521102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800187892.hg.1 | NR_109954.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800187892.hg.1 | TC0800012449.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0800012449.hg.1 | ENST00000500902.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012449.hg.1 | ENST00000500902.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012449.hg.1 | ENST00000500902.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012449.hg.1 | ENST00000500902.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012449.hg.1 | ENST00000521102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012449.hg.1 | ENST00000521102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012449.hg.1 | ENST00000521102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012449.hg.1 | NR_109954.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012449.hg.1 | NR_109954.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012449.hg.1 | NR_109954.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012449.hg.1 | NR_109954.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800187894.hg.1 | ENST00000500902.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800187894.hg.1 | NR_109954.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800187894.hg.1 | ENST00000500902.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800187894.hg.1 | NR_109954.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800187894.hg.1 | TC0800012449.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800187890.hg.1 | ENST00000521102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800187890.hg.1 | NR_109954.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800187890.hg.1 | ENST00000521102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800187890.hg.1 | NR_109954.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800187890.hg.1 | TC0800012449.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800187893.hg.1 | ENST00000500902.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800187893.hg.1 | ENST00000521102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800187893.hg.1 | NR_109954.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800187893.hg.1 | ENST00000500902.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800187893.hg.1 | ENST00000521102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800187893.hg.1 | NR_109954.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800187893.hg.1 | TC0800012449.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A036029 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A036029 | - | - |

BAALC-AS1
Genomic Map