Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027723.hg.1 |
ENST00000499809.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027723.hg.1 |
ENST00000514294.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027723.hg.1 |
ENST00000535683.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027723.hg.1 |
ENST00000537070.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027723.hg.1 |
NR_034148.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027723.hg.1 |
TC11002319.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012401.hg.1 |
ENST00000499809.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012401.hg.1 |
ENST00000499809.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012401.hg.1 |
ENST00000499809.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012401.hg.1 |
ENST00000499809.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012401.hg.1 |
ENST00000499809.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012401.hg.1 |
ENST00000499809.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012401.hg.1 |
ENST00000499809.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012401.hg.1 |
ENST00000499809.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012401.hg.1 |
ENST00000501294.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012401.hg.1 |
ENST00000501294.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012401.hg.1 |
ENST00000501294.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012401.hg.1 |
ENST00000501294.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012401.hg.1 |
ENST00000501294.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012401.hg.1 |
ENST00000501294.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012401.hg.1 |
ENST00000501294.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012401.hg.1 |
ENST00000501294.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012401.hg.1 |
ENST00000514294.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012401.hg.1 |
ENST00000514294.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012401.hg.1 |
ENST00000514294.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012401.hg.1 |
ENST00000514294.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012401.hg.1 |
ENST00000514294.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012401.hg.1 |
ENST00000514294.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012401.hg.1 |
ENST00000514294.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012401.hg.1 |
ENST00000535683.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012401.hg.1 |
ENST00000535683.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012401.hg.1 |
ENST00000535683.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012401.hg.1 |
ENST00000535683.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012401.hg.1 |
ENST00000537070.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012401.hg.1 |
ENST00000537070.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012401.hg.1 |
ENST00000537070.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012401.hg.1 |
ENST00000537070.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012401.hg.1 |
ENST00000537070.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012401.hg.1 |
ENST00000602803.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012401.hg.1 |
ENST00000602803.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012401.hg.1 |
NR_034148.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012401.hg.1 |
NR_034148.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012401.hg.1 |
NR_034148.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012401.hg.1 |
NR_034148.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012401.hg.1 |
NR_034148.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012401.hg.1 |
NR_034148.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012401.hg.1 |
NR_034148.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027720.hg.1 |
ENST00000499809.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027720.hg.1 |
ENST00000501294.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027720.hg.1 |
ENST00000514294.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027720.hg.1 |
ENST00000535683.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027720.hg.1 |
ENST00000537070.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027720.hg.1 |
NR_034148.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027720.hg.1 |
TC11002319.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100195238.hg.1 |
ENST00000499809.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195238.hg.1 |
ENST00000501294.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195238.hg.1 |
ENST00000514294.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195238.hg.1 |
ENST00000535683.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195238.hg.1 |
ENST00000537070.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195238.hg.1 |
NR_034148.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100195238.hg.1 |
ENST00000499809.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195238.hg.1 |
ENST00000501294.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195238.hg.1 |
ENST00000514294.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195238.hg.1 |
ENST00000535683.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195238.hg.1 |
ENST00000537070.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195238.hg.1 |
NR_034148.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100195238.hg.1 |
TC1100012401.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100195237.hg.1 |
ENST00000499809.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195237.hg.1 |
ENST00000501294.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195237.hg.1 |
ENST00000514294.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195237.hg.1 |
ENST00000535683.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195237.hg.1 |
ENST00000537070.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195237.hg.1 |
NR_034148.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100195237.hg.1 |
ENST00000499809.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195237.hg.1 |
ENST00000501294.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195237.hg.1 |
ENST00000514294.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195237.hg.1 |
ENST00000535683.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195237.hg.1 |
ENST00000537070.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195237.hg.1 |
NR_034148.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100195237.hg.1 |
TC1100012401.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100012401.hg.1 |
ENST00000499809.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012401.hg.1 |
ENST00000499809.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012401.hg.1 |
ENST00000499809.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012401.hg.1 |
ENST00000499809.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012401.hg.1 |
ENST00000499809.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012401.hg.1 |
ENST00000499809.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012401.hg.1 |
ENST00000499809.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012401.hg.1 |
ENST00000499809.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012401.hg.1 |
ENST00000501294.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012401.hg.1 |
ENST00000501294.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012401.hg.1 |
ENST00000501294.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012401.hg.1 |
ENST00000501294.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012401.hg.1 |
ENST00000501294.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012401.hg.1 |
ENST00000501294.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012401.hg.1 |
ENST00000501294.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012401.hg.1 |
ENST00000501294.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012401.hg.1 |
ENST00000514294.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012401.hg.1 |
ENST00000514294.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012401.hg.1 |
ENST00000514294.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012401.hg.1 |
ENST00000514294.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012401.hg.1 |
ENST00000514294.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012401.hg.1 |
ENST00000514294.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012401.hg.1 |
ENST00000514294.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012401.hg.1 |
ENST00000535683.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012401.hg.1 |
ENST00000535683.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012401.hg.1 |
ENST00000535683.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012401.hg.1 |
ENST00000535683.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012401.hg.1 |
ENST00000537070.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012401.hg.1 |
ENST00000537070.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012401.hg.1 |
ENST00000537070.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012401.hg.1 |
ENST00000537070.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012401.hg.1 |
ENST00000537070.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012401.hg.1 |
ENST00000602803.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012401.hg.1 |
ENST00000602803.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012401.hg.1 |
NR_034148.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012401.hg.1 |
NR_034148.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012401.hg.1 |
NR_034148.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012401.hg.1 |
NR_034148.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012401.hg.1 |
NR_034148.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012401.hg.1 |
NR_034148.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012401.hg.1 |
NR_034148.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100195243.hg.1 |
ENST00000499809.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195243.hg.1 |
ENST00000514294.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195243.hg.1 |
ENST00000535683.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195243.hg.1 |
ENST00000537070.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195243.hg.1 |
NR_034148.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100195243.hg.1 |
ENST00000499809.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195243.hg.1 |
ENST00000514294.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195243.hg.1 |
ENST00000535683.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195243.hg.1 |
ENST00000537070.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195243.hg.1 |
NR_034148.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100195243.hg.1 |
TC1100012401.hg.1 |
Clariom D TC |