| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13008224.hg.1 |
NR_036128.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13008224.hg.1 |
TC13000706.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009111.hg.1 |
ENST00000578032.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009111.hg.1 |
NR_036128.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300161461.hg.1 |
ENST00000578032.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300161461.hg.1 |
NR_036128.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300161461.hg.1 |
ENST00000578032.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300161461.hg.1 |
NR_036128.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300161461.hg.1 |
TC1300009111.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1300009111.hg.1 |
ENST00000578032.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009111.hg.1 |
NR_036128.1 |
Refseq |