Assay Details
| Gene Symbol: ZBTB20-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038993.1 | 6 - 7 | 933 | 103 | ||
| GenBank mRNA | AK098660.1 | - | 6 - 7 | 909 | 103 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 568 | MIR568 |
MIRN568, hsa-mir-568 |
-50.4kb | Anti-sense | Chr. 3 - 114316475 - 114316569 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC101929754 | LOC101929754 | 77.8kb | Sense | Chr. 3 - 114453593 - 114520132 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03000589.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03010820.hg.1 | TC03000589.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03010820.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03010820.hg.1 | NR_038993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03010818.hg.1 | TC03000589.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03010818.hg.1 | ENST00000460210.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03010818.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03010818.hg.1 | NR_038993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000460210.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000460210.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000460210.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000460210.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000467304.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000467304.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000467304.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000467304.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000467304.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000467304.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000467304.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000487814.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000487814.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000496219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000496219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000496219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000496219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000496219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000496219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000496219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000496219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000496219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000496219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000496219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000496219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | NR_038993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | NR_038993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | NR_038993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | NR_038993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | NR_038993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | NR_038993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | NR_038993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | NR_038993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | NR_038993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | NR_038993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | NR_038993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | NR_038993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | NR_038993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | NR_038993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03010817.hg.1 | TC03000589.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03010817.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03010817.hg.1 | NR_038993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03010816.hg.1 | TC03000589.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03010816.hg.1 | ENST00000467304.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03010816.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03010816.hg.1 | NR_038993.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300159982.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300159982.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300159982.hg.1 | NR_038993.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300159982.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300159982.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300159982.hg.1 | NR_038993.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300159982.hg.1 | TC0300008389.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300159978.hg.1 | ENST00000467304.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300159978.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300159978.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300159978.hg.1 | NR_038993.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300159978.hg.1 | ENST00000467304.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300159978.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300159978.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300159978.hg.1 | NR_038993.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300159978.hg.1 | TC0300008389.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300159979.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300159979.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300159979.hg.1 | NR_038993.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300159979.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300159979.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300159979.hg.1 | NR_038993.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300159979.hg.1 | TC0300008389.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300159980.hg.1 | ENST00000460210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300159980.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300159980.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300159980.hg.1 | NR_038993.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300159980.hg.1 | ENST00000460210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300159980.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300159980.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300159980.hg.1 | NR_038993.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300159980.hg.1 | TC0300008389.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000460210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000460210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000460210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000460210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000467304.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000467304.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000467304.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000467304.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000467304.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000467304.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000467304.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000487814.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000487814.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000496219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000496219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000496219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000496219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000496219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000496219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000496219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000496219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000496219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000496219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000496219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000496219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | NR_038993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | NR_038993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | NR_038993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | NR_038993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | NR_038993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | NR_038993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | NR_038993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | NR_038993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | NR_038993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | NR_038993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | NR_038993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | NR_038993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | NR_038993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | NR_038993.1 | Refseq |

ZBTB20-AS1
Genomic Map