Assay Details
| Gene Symbol: ZBTB20-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038993.1 | 8 - 9 | 1136 | 84 | ||
| GenBank mRNA | AK098660.1 | - | 8 - 9 | 1112 | 84 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 568 | MIR568 |
MIRN568, hsa-mir-568 |
-70.9kb | Anti-sense | Chr. 3 - 114316475 - 114316569 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC101929754 | LOC101929754 | 64.6kb | Sense | Chr. 3 - 114453593 - 114520132 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03000589.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P728215 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 14490 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P728215 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P728215 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 231497_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000460210.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000460210.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000460210.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000460210.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000467304.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000467304.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000467304.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000467304.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000467304.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000467304.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000467304.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000487814.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000487814.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000496219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000496219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000496219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000496219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000496219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000496219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000496219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000496219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000496219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000496219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000496219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | ENST00000496219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | NR_038993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | NR_038993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | NR_038993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | NR_038993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | NR_038993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | NR_038993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | NR_038993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | NR_038993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | NR_038993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | NR_038993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | NR_038993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | NR_038993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | NR_038993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008389.hg.1 | NR_038993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03010822.hg.1 | TC03000589.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03010822.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03010822.hg.1 | ENST00000487814.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03010822.hg.1 | NR_038993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000460210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000460210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000460210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000460210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000467304.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000467304.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000467304.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000467304.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000467304.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000467304.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000467304.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000487814.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000487814.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000496219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000496219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000496219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000496219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000496219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000496219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000496219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000496219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000496219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000496219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000496219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | ENST00000496219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | NR_038993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | NR_038993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | NR_038993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | NR_038993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | NR_038993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | NR_038993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | NR_038993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | NR_038993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | NR_038993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | NR_038993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | NR_038993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | NR_038993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | NR_038993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008389.hg.1 | NR_038993.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300159986.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300159986.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300159986.hg.1 | ENST00000487814.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300159986.hg.1 | NR_038993.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300159986.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300159986.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300159986.hg.1 | ENST00000487814.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300159986.hg.1 | NR_038993.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300159986.hg.1 | TC0300008389.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300159983.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300159983.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300159983.hg.1 | NR_038993.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300159983.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300159983.hg.1 | ENST00000475939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300159983.hg.1 | NR_038993.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300159983.hg.1 | TC0300008389.hg.1 | Clariom D TC |
| Human Genome U133 Plus 2.0 Array | 231497_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 231497_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 231497_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 231497_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 231497_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 231497_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 231497_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 231497_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 231497_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 231497_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 231497_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 231497_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 231497_at10 | - | - |
| Human Genome U95 Set | 76859_at | - | - |
| Human Genome U95 Set | 76859_PM_at | - | - |
| Human Genome U95 Set | 87946_at | - | - |
| Human Genome U95 Set | 87946_PM_at | - | - |
| Human Genome U133 Set | 231497_at | - | - |
| Human Genome U133 Set | 231497_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A023022 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A023022 | - | - |

ZBTB20-AS1
Genomic Map