Assay Details
| Gene Symbol: MIR378F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_039615.1 | 1 | 37 | 61 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cannabinoid receptor 2 | CNR2 |
CB-2, CB2, CX5 |
-15.7kb | Anti-sense | Chr. 1 - 23870526 - 23913327 on NCBI Build GRCh38 |
| Downstream | proline rich nuclear receptor coactivator 2 | PNRC2 | 30.7kb | Sense | Chr. 1 - 23959164 - 23963462 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC0100007341.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01000304.hg.1
Set Membership: |
|

MIR378F
Genomic Map