Assay Details
Gene Symbol: MIR4500 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_039722.1 | 1 | 34 | 69 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | MIR4500 host gene | MIR4500HG | Sense | Chr. 13 - 87443987 - 87670963 on NCBI Build GRCh38 | ||
Downstream | SLIT and NTRK like family member 5 | SLITRK5 |
LRRC11, bA364G4.2 |
52.5kb | Anti-sense | Chr. 13 - 87671328 - 87679619 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1300009407.hg.1
This assay targets the same location as HTA transcript cluster(s) TC13000772.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
