Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008111.hg.1 |
ENST00000584082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008111.hg.1 |
NR_039740.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15007246.hg.1 |
NR_039740.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15007246.hg.1 |
TC15000795.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1500008111.hg.1 |
ENST00000584082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008111.hg.1 |
NR_039740.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500158339.hg.1 |
ENST00000584082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500158339.hg.1 |
NR_039740.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500158339.hg.1 |
ENST00000584082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500158339.hg.1 |
NR_039740.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500158339.hg.1 |
TC1500008111.hg.1 |
Clariom D TC |