Assay Details
| Gene Symbol: MIR3978 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_039774.1 | 1 | 47 | 85 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 652 | MIR652 |
MIRN652, hsa-mir-652 |
-26.7kb | Sense | Chr. X - 110055329 - 110055426 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC101928589 | LOC101928589 | 93.6kb | Sense | Chr. X - 110175773 - 110177788 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC0X00008113.hg.1
This assay targets the same location as HTA transcript cluster(s) TC0X000540.hg.1
Set Membership: |
|

MIR3978
Genomic Map