Assay Details
| Gene Symbol: MIR4660 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_039804.1 | 1 | 39 | 62 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | exoribonuclease 1 | ERI1 |
3'HEXO, HEXO, THEX1 |
Sense | Chr. 8 - 8892225 - 9100262 on NCBI Build GRCh38 | |
| Downstream | protein phosphatase 1 regulatory subunit 3B | PPP1R3B |
GL, PPP1R4, PTG |
87.7kb | Anti-sense | Chr. 8 - 9136254 - 9151642 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC0800006664.hg.1
This assay targets the same location as HTA transcript cluster(s) TC08000064.hg.1
Set Membership: |
|

MIR4660
Genomic Map