Assay Details
| Gene Symbol: MIR4677 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_039824.1 | 1 | 38 | 70 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | serologically defined colon cancer antigen 8 | SDCCAG8 |
BBS16, CCCAP, CCCAP SLSN7, HSPC085, NPHP10, NY-CO-8, SLSN7, hCCCAP |
Sense | Chr. 1 - 243255328 - 243500092 on NCBI Build GRCh38 | |
| Downstream | AKT serine/threonine kinase 3 | AKT3 |
MPPH, MPPH2, PKB-GAMMA, PKBG, PRKBG, RAC-PK-gamma, RAC-gamma, STK-2 |
142.0kb | Anti-sense | Chr. 1 - 243488233 - 243851079 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC0100012202.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01001982.hg.1
Set Membership: |
|

MIR4677
Genomic Map