Assay Details
| Gene Symbol: MIR4682 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_039830.1 | 1 | 37 | 70 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | SEC23 interacting protein | SEC23IP |
MSTP053, P125, P125A |
-13.9kb | Sense | Chr. 10 - 119892573 - 119944658 on NCBI Build GRCh38 |
| Downstream | phospholipid phosphatase 4 | PLPP4 |
DPPL2, PPAPDC1, PPAPDC1A |
498.4kb | Sense | Chr. 10 - 120456954 - 120600123 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC1000009097.hg.1
This assay targets the same location as HTA transcript cluster(s) TC10000872.hg.1
Set Membership: |
|

MIR4682
Genomic Map