Assay Details
| Gene Symbol: MIR4696 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_039845.1 | 1 | 40 | 64 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | chordin like 2 | CHRDL2 |
BNF1, CHL2 |
Sense | Chr. 11 - 74696428 - 74731385 on NCBI Build GRCh38 | |
| Downstream | ring finger protein 169 | RNF169 | 28.5kb | Anti-sense | Chr. 11 - 74748856 - 74842413 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC1100011625.hg.1
This assay targets the same location as HTA transcript cluster(s) TC11002086.hg.1
Set Membership: |
|

MIR4696
Genomic Map