| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16009036.hg.1 |
NR_039871.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16009036.hg.1 |
TC16000639.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008580.hg.1 |
ENST00000563750.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008580.hg.1 |
ENST00000563750.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008580.hg.1 |
ENST00000567335.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008580.hg.1 |
ENST00000567335.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008580.hg.1 |
ENST00000568107.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008580.hg.1 |
ENST00000568107.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008580.hg.1 |
ENST00000568107.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008580.hg.1 |
ENST00000568107.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008580.hg.1 |
ENST00000568107.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008580.hg.1 |
ENST00000568107.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008580.hg.1 |
ENST00000568107.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008580.hg.1 |
ENST00000568107.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008580.hg.1 |
ENST00000568107.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008580.hg.1 |
ENST00000568107.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008580.hg.1 |
ENST00000568107.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008580.hg.1 |
ENST00000568107.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008580.hg.1 |
ENST00000568107.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008580.hg.1 |
ENST00000568107.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008580.hg.1 |
ENST00000568107.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008580.hg.1 |
ENST00000568107.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008580.hg.1 |
ENST00000568107.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008580.hg.1 |
ENST00000568107.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008580.hg.1 |
ENST00000568107.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008580.hg.1 |
ENST00000568107.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008580.hg.1 |
ENST00000568107.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008580.hg.1 |
ENST00000568107.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008580.hg.1 |
ENST00000568107.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008580.hg.1 |
ENST00000568107.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008580.hg.1 |
ENST00000568107.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008580.hg.1 |
ENST00000568107.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008580.hg.1 |
ENST00000568107.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008580.hg.1 |
ENST00000568107.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008580.hg.1 |
ENST00000568107.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008580.hg.1 |
ENST00000581455.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008580.hg.1 |
NM_022041.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008580.hg.1 |
NM_022041.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008580.hg.1 |
NM_022041.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008580.hg.1 |
NM_022041.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008580.hg.1 |
NM_022041.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008580.hg.1 |
NM_022041.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008580.hg.1 |
NM_022041.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008580.hg.1 |
NM_022041.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008580.hg.1 |
NM_022041.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008580.hg.1 |
NM_022041.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008580.hg.1 |
NM_022041.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008580.hg.1 |
NM_022041.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008580.hg.1 |
NM_022041.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008580.hg.1 |
NM_022041.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008580.hg.1 |
NM_022041.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008580.hg.1 |
NR_039871.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008580.hg.1 |
ENST00000563750.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008580.hg.1 |
ENST00000563750.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008580.hg.1 |
ENST00000567335.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008580.hg.1 |
ENST00000567335.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008580.hg.1 |
ENST00000568107.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008580.hg.1 |
ENST00000568107.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008580.hg.1 |
ENST00000568107.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008580.hg.1 |
ENST00000568107.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008580.hg.1 |
ENST00000568107.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008580.hg.1 |
ENST00000568107.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008580.hg.1 |
ENST00000568107.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008580.hg.1 |
ENST00000568107.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008580.hg.1 |
ENST00000568107.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008580.hg.1 |
ENST00000568107.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008580.hg.1 |
ENST00000568107.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008580.hg.1 |
ENST00000568107.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008580.hg.1 |
ENST00000568107.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008580.hg.1 |
ENST00000568107.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008580.hg.1 |
ENST00000568107.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008580.hg.1 |
ENST00000568107.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008580.hg.1 |
ENST00000568107.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008580.hg.1 |
ENST00000568107.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008580.hg.1 |
ENST00000568107.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008580.hg.1 |
ENST00000568107.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008580.hg.1 |
ENST00000568107.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008580.hg.1 |
ENST00000568107.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008580.hg.1 |
ENST00000568107.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008580.hg.1 |
ENST00000568107.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008580.hg.1 |
ENST00000568107.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008580.hg.1 |
ENST00000568107.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008580.hg.1 |
ENST00000568107.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008580.hg.1 |
ENST00000568107.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008580.hg.1 |
ENST00000568107.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008580.hg.1 |
ENST00000581455.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008580.hg.1 |
NM_022041.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008580.hg.1 |
NM_022041.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008580.hg.1 |
NM_022041.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008580.hg.1 |
NM_022041.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008580.hg.1 |
NM_022041.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008580.hg.1 |
NM_022041.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008580.hg.1 |
NM_022041.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008580.hg.1 |
NM_022041.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008580.hg.1 |
NM_022041.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008580.hg.1 |
NM_022041.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008580.hg.1 |
NM_022041.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008580.hg.1 |
NM_022041.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008580.hg.1 |
NM_022041.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008580.hg.1 |
NM_022041.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008580.hg.1 |
NM_022041.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008580.hg.1 |
NR_039871.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600164046.hg.1 |
ENST00000568107.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600164046.hg.1 |
ENST00000581455.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600164046.hg.1 |
NR_039871.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600164046.hg.1 |
ENST00000568107.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600164046.hg.1 |
ENST00000581455.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600164046.hg.1 |
NR_039871.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600164046.hg.1 |
TC1600008580.hg.1 |
Clariom D TC |