| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19025958.hg.1 |
NR_039910.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19025958.hg.1 |
TC19001916.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011623.hg.1 |
ENST00000582477.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011623.hg.1 |
NR_039910.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900198372.hg.1 |
ENST00000582477.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900198372.hg.1 |
NR_039910.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900198372.hg.1 |
TC1900011623.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900198372.hg.1 |
ENST00000582477.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900198372.hg.1 |
NR_039910.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011623.hg.1 |
ENST00000582477.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011623.hg.1 |
NR_039910.1 |
Refseq |