Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02039450.hg.1 |
TC02002495.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02039450.hg.1 |
ENST00000489714.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02039450.hg.1 |
NR_045375.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02039449.hg.1 |
TC02002495.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02039449.hg.1 |
ENST00000489714.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02039449.hg.1 |
ENST00000536175.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02039449.hg.1 |
NR_045375.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02039454.hg.1 |
TC02002495.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02039454.hg.1 |
ENST00000536175.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02039454.hg.1 |
NR_045375.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02039453.hg.1 |
TC02002495.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02039453.hg.1 |
ENST00000489714.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02039453.hg.1 |
ENST00000536175.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02039453.hg.1 |
NR_045375.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02039456.hg.1 |
TC02002495.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02039456.hg.1 |
NR_045375.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016746.hg.1 |
ENST00000486672.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016746.hg.1 |
ENST00000486672.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016746.hg.1 |
ENST00000486672.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016746.hg.1 |
ENST00000486672.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016746.hg.1 |
ENST00000486672.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016746.hg.1 |
ENST00000489714.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016746.hg.1 |
ENST00000489714.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016746.hg.1 |
ENST00000489714.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016746.hg.1 |
ENST00000489714.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016746.hg.1 |
NR_045375.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016746.hg.1 |
NR_045375.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016746.hg.1 |
NR_045375.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016746.hg.1 |
NR_045375.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016746.hg.1 |
NR_045375.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016746.hg.1 |
NR_045375.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016746.hg.1 |
NR_045375.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02039455.hg.1 |
TC02002495.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02039455.hg.1 |
NR_045375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200227998.hg.1 |
ENST00000486672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200227998.hg.1 |
ENST00000489714.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200227998.hg.1 |
NR_045375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200227998.hg.1 |
ENST00000486672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200227998.hg.1 |
ENST00000489714.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200227998.hg.1 |
NR_045375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200227998.hg.1 |
TC0200016746.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200228000.hg.1 |
ENST00000486672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200228000.hg.1 |
NR_045375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200228000.hg.1 |
ENST00000486672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200228000.hg.1 |
NR_045375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200228000.hg.1 |
TC0200016746.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200227999.hg.1 |
ENST00000486672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200227999.hg.1 |
NR_045375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200227999.hg.1 |
ENST00000486672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200227999.hg.1 |
NR_045375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200227999.hg.1 |
TC0200016746.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200227995.hg.1 |
ENST00000489714.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200227995.hg.1 |
NR_045375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200227995.hg.1 |
ENST00000489714.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200227995.hg.1 |
NR_045375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200227995.hg.1 |
TC0200016746.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200227994.hg.1 |
ENST00000486672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200227994.hg.1 |
ENST00000489714.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200227994.hg.1 |
NR_045375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200227994.hg.1 |
ENST00000486672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200227994.hg.1 |
ENST00000489714.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200227994.hg.1 |
NR_045375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200227994.hg.1 |
TC0200016746.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200228001.hg.1 |
NR_045375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200228001.hg.1 |
NR_045375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200228001.hg.1 |
TC0200016746.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200016746.hg.1 |
ENST00000486672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016746.hg.1 |
ENST00000486672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016746.hg.1 |
ENST00000486672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016746.hg.1 |
ENST00000486672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016746.hg.1 |
ENST00000486672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016746.hg.1 |
ENST00000489714.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016746.hg.1 |
ENST00000489714.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016746.hg.1 |
ENST00000489714.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016746.hg.1 |
ENST00000489714.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016746.hg.1 |
NR_045375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016746.hg.1 |
NR_045375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016746.hg.1 |
NR_045375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016746.hg.1 |
NR_045375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016746.hg.1 |
NR_045375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016746.hg.1 |
NR_045375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016746.hg.1 |
NR_045375.2 |
Refseq |