Assay Details
Gene Symbol: FAAHP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045483.1 | 6 - 7 | 506 | 99 | ||
GenBank mRNA | BI756183.1 | - | 3 - 4 | 197 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
