Assay Details
| Gene Symbol: LOC100129138 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033990.1 | 1 | 623 | 84 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
amylase, alpha 1C (salivary) |
AMY1C | AMY1 | -314.3kb | Sense | Chr. 1 - 103745430 - 103758692 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC101928476 | LOC101928476 | 1.5mb | Anti-sense | Chr. 1 - 105589694 - 105618935 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000922.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15093913 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20459235 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20459231 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20459241 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20459242 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20459238 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00428026 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20459226 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00428016 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15093902 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15093912 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00428028 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20459253 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20459178 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00428043 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20459250 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00428004 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00428029 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20459179 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20459211 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20459259 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20459195 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20459197 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20459182 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20459267 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20459224 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20459212 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00428064 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20459280 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20459183 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20459209 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20459191 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20459201 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00427971 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20459200 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20459181 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20459254 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20459233 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20459223 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20459266 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00428006 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20459186 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00428054 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20459281 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00428000 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20459258 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00428042 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20459230 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00427976 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20459225 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20459243 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20459268 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20459277 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20459261 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20459285 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00427993 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20459244 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20459284 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20459257 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20459177 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20459194 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20459247 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20459283 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20459234 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20459175 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00427972 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00427991 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00428017 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20459180 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20459271 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00428001 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20459256 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20459275 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20459264 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20459199 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20459278 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20459251 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20459255 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20459205 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20459202 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20459187 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20459269 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20459210 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20459192 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20459239 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20459203 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20459248 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00428053 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00428060 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 614172 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 39350 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P280833 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 25364 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 44987 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P280833 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P280833 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P35252579 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P15242503 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P35252584 | - | - |
| Agilent Human 1B Microarray G4111A | A_32_P910283 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P15242503 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01014485.hg.1 | TC01000922.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014485.hg.1 | ENST00000418362.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014485.hg.1 | NR_033990.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018250.hg.1 | ENST00000401022.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018250.hg.1 | ENST00000418362.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018250.hg.1 | ENST00000418362.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018250.hg.1 | ENST00000418362.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018250.hg.1 | NR_033990.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018250.hg.1 | NR_033990.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018250.hg.1 | NR_033990.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014486.hg.1 | TC01000922.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014486.hg.1 | ENST00000418362.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014486.hg.1 | NR_033990.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100238459.hg.1 | ENST00000401022.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100238459.hg.1 | ENST00000418362.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100238459.hg.1 | NR_033990.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100238459.hg.1 | ENST00000401022.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100238459.hg.1 | ENST00000418362.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100238459.hg.1 | NR_033990.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100238459.hg.1 | TC0100018250.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100238460.hg.1 | ENST00000418362.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100238460.hg.1 | NR_033990.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100238460.hg.1 | ENST00000418362.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100238460.hg.1 | NR_033990.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100238460.hg.1 | TC0100018250.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100238461.hg.1 | ENST00000418362.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100238461.hg.1 | NR_033990.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100238461.hg.1 | ENST00000418362.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100238461.hg.1 | NR_033990.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100238461.hg.1 | TC0100018250.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100018250.hg.1 | ENST00000401022.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018250.hg.1 | ENST00000418362.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018250.hg.1 | ENST00000418362.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018250.hg.1 | ENST00000418362.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018250.hg.1 | NR_033990.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018250.hg.1 | NR_033990.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018250.hg.1 | NR_033990.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A026994 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A026994 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 234887 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3258046 | NR_033990.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3258046 | BC014218.2 | human_transcriptome |

LOC100129138
Genomic Map