| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01039070.hg.1 |
TC01002516.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01039070.hg.1 |
ENST00000424989.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01039070.hg.1 |
NR_038842.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01039073.hg.1 |
TC01002516.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01039073.hg.1 |
NR_038842.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013741.hg.1 |
ENST00000424989.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013741.hg.1 |
ENST00000424989.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013741.hg.1 |
ENST00000424989.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013741.hg.1 |
ENST00000424989.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013741.hg.1 |
ENST00000424989.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013741.hg.1 |
NR_038842.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013741.hg.1 |
NR_038842.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013741.hg.1 |
NR_038842.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013741.hg.1 |
NR_038842.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013741.hg.1 |
NR_038842.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013741.hg.1 |
NR_038842.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01039071.hg.1 |
TC01002516.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01039071.hg.1 |
ENST00000424989.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01039071.hg.1 |
NR_038842.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01039072.hg.1 |
TC01002516.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01039072.hg.1 |
ENST00000424989.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01039072.hg.1 |
NR_038842.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100202448.hg.1 |
ENST00000424989.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100202448.hg.1 |
NR_038842.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100202448.hg.1 |
ENST00000424989.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100202448.hg.1 |
NR_038842.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100202448.hg.1 |
TC0100013741.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100202452.hg.1 |
ENST00000424989.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100202452.hg.1 |
NR_038842.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100202452.hg.1 |
ENST00000424989.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100202452.hg.1 |
NR_038842.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100202452.hg.1 |
TC0100013741.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100202449.hg.1 |
ENST00000424989.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100202449.hg.1 |
NR_038842.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100202449.hg.1 |
ENST00000424989.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100202449.hg.1 |
NR_038842.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100202449.hg.1 |
TC0100013741.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100013741.hg.1 |
ENST00000424989.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013741.hg.1 |
ENST00000424989.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013741.hg.1 |
ENST00000424989.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013741.hg.1 |
ENST00000424989.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013741.hg.1 |
ENST00000424989.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013741.hg.1 |
NR_038842.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013741.hg.1 |
NR_038842.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013741.hg.1 |
NR_038842.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013741.hg.1 |
NR_038842.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013741.hg.1 |
NR_038842.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013741.hg.1 |
NR_038842.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100202454.hg.1 |
NR_038842.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100202454.hg.1 |
NR_038842.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100202454.hg.1 |
TC0100013741.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100202450.hg.1 |
ENST00000424989.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100202450.hg.1 |
NR_038842.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100202450.hg.1 |
ENST00000424989.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100202450.hg.1 |
NR_038842.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100202450.hg.1 |
TC0100013741.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100202453.hg.1 |
ENST00000424989.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100202453.hg.1 |
NR_038842.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100202453.hg.1 |
ENST00000424989.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100202453.hg.1 |
NR_038842.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100202453.hg.1 |
TC0100013741.hg.1 |
Clariom D TC |