Assay Details
| Gene Symbol: NUDT16L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193452.1 | NP_001180381.1 | 2 - 3 | 516 | 79 | |
| GenBank mRNA | BQ923183.1 | - | 2 - 3 | 500 | 79 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16000134.hg.1
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600006752.hg.1 | ENST00000304301.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006752.hg.1 | ENST00000304301.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006752.hg.1 | ENST00000304301.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006752.hg.1 | ENST00000304301.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006752.hg.1 | ENST00000304301.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006752.hg.1 | ENST00000304301.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006752.hg.1 | ENST00000304301.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006752.hg.1 | ENST00000304301.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006752.hg.1 | ENST00000304301.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006752.hg.1 | ENST00000405142.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006752.hg.1 | ENST00000405142.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006752.hg.1 | ENST00000405142.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006752.hg.1 | ENST00000405142.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006752.hg.1 | ENST00000405142.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006752.hg.1 | ENST00000405142.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006752.hg.1 | ENST00000405142.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006752.hg.1 | ENST00000405142.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006752.hg.1 | ENST00000405142.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006752.hg.1 | ENST00000405142.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006752.hg.1 | ENST00000405142.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006752.hg.1 | ENST00000586252.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006752.hg.1 | ENST00000586252.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006752.hg.1 | ENST00000586252.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006752.hg.1 | ENST00000586252.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006752.hg.1 | ENST00000586252.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006752.hg.1 | ENST00000586252.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006752.hg.1 | ENST00000586536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006752.hg.1 | ENST00000586536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006752.hg.1 | ENST00000586536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006752.hg.1 | ENST00000586536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006752.hg.1 | ENST00000586536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006752.hg.1 | ENST00000586536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006752.hg.1 | ENST00000586536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006752.hg.1 | ENST00000586536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006752.hg.1 | ENST00000586536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006752.hg.1 | ENST00000586536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006752.hg.1 | ENST00000590460.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006752.hg.1 | ENST00000590460.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006752.hg.1 | ENST00000590460.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006752.hg.1 | ENST00000590460.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006752.hg.1 | ENST00000590460.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006752.hg.1 | ENST00000590460.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006752.hg.1 | NM_001193452.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006752.hg.1 | NM_001193452.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006752.hg.1 | NM_001193452.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006752.hg.1 | NM_001193452.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006752.hg.1 | NM_001193452.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006752.hg.1 | NM_001193452.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006752.hg.1 | NM_001193452.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006752.hg.1 | NM_001193452.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006752.hg.1 | NM_001193452.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006752.hg.1 | NM_001193452.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006752.hg.1 | NM_032349.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006752.hg.1 | NM_032349.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006752.hg.1 | NM_032349.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006752.hg.1 | NM_032349.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006752.hg.1 | NM_032349.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006752.hg.1 | NM_032349.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006752.hg.1 | NM_032349.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006752.hg.1 | NM_032349.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006752.hg.1 | NM_032349.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16002117.hg.1 | ENST00000405142.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16002117.hg.1 | NM_001193452.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16002117.hg.1 | TC16000134.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1600148195.hg.1 | ENST00000405142.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600148195.hg.1 | ENST00000586536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600148195.hg.1 | NM_001193452.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600148195.hg.1 | ENST00000405142.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600148195.hg.1 | ENST00000586536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600148195.hg.1 | NM_001193452.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600148195.hg.1 | TC1600006752.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600148202.hg.1 | ENST00000304301.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600148202.hg.1 | ENST00000405142.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600148202.hg.1 | ENST00000586536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600148202.hg.1 | NM_001193452.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600148202.hg.1 | NM_032349.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600148202.hg.1 | ENST00000304301.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600148202.hg.1 | ENST00000405142.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600148202.hg.1 | ENST00000586536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600148202.hg.1 | NM_001193452.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600148202.hg.1 | NM_032349.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600148202.hg.1 | TC1600006752.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1600006752.hg.1 | ENST00000304301.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006752.hg.1 | ENST00000304301.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006752.hg.1 | ENST00000304301.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006752.hg.1 | ENST00000304301.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006752.hg.1 | ENST00000304301.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006752.hg.1 | ENST00000304301.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006752.hg.1 | ENST00000304301.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006752.hg.1 | ENST00000304301.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006752.hg.1 | ENST00000304301.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006752.hg.1 | ENST00000405142.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006752.hg.1 | ENST00000405142.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006752.hg.1 | ENST00000405142.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006752.hg.1 | ENST00000405142.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006752.hg.1 | ENST00000405142.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006752.hg.1 | ENST00000405142.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006752.hg.1 | ENST00000405142.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006752.hg.1 | ENST00000405142.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006752.hg.1 | ENST00000405142.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006752.hg.1 | ENST00000405142.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006752.hg.1 | ENST00000405142.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006752.hg.1 | ENST00000586252.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006752.hg.1 | ENST00000586252.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006752.hg.1 | ENST00000586252.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006752.hg.1 | ENST00000586252.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006752.hg.1 | ENST00000586252.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006752.hg.1 | ENST00000586252.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006752.hg.1 | ENST00000586536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006752.hg.1 | ENST00000586536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006752.hg.1 | ENST00000586536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006752.hg.1 | ENST00000586536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006752.hg.1 | ENST00000586536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006752.hg.1 | ENST00000586536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006752.hg.1 | ENST00000586536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006752.hg.1 | ENST00000586536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006752.hg.1 | ENST00000586536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006752.hg.1 | ENST00000586536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006752.hg.1 | ENST00000590460.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006752.hg.1 | ENST00000590460.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006752.hg.1 | ENST00000590460.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006752.hg.1 | ENST00000590460.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006752.hg.1 | ENST00000590460.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006752.hg.1 | ENST00000590460.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006752.hg.1 | NM_001193452.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006752.hg.1 | NM_001193452.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006752.hg.1 | NM_001193452.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006752.hg.1 | NM_001193452.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006752.hg.1 | NM_001193452.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006752.hg.1 | NM_001193452.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006752.hg.1 | NM_001193452.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006752.hg.1 | NM_001193452.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006752.hg.1 | NM_001193452.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006752.hg.1 | NM_001193452.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006752.hg.1 | NM_032349.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006752.hg.1 | NM_032349.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006752.hg.1 | NM_032349.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006752.hg.1 | NM_032349.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006752.hg.1 | NM_032349.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006752.hg.1 | NM_032349.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006752.hg.1 | NM_032349.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006752.hg.1 | NM_032349.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006752.hg.1 | NM_032349.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600148198.hg.1 | ENST00000304301.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600148198.hg.1 | ENST00000405142.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600148198.hg.1 | ENST00000586536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600148198.hg.1 | ENST00000590460.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600148198.hg.1 | NM_001193452.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600148198.hg.1 | NM_032349.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600148198.hg.1 | ENST00000304301.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600148198.hg.1 | ENST00000405142.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600148198.hg.1 | ENST00000586536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600148198.hg.1 | ENST00000590460.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600148198.hg.1 | NM_001193452.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600148198.hg.1 | NM_032349.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600148198.hg.1 | TC1600006752.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600148199.hg.1 | ENST00000304301.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600148199.hg.1 | ENST00000405142.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600148199.hg.1 | ENST00000586252.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600148199.hg.1 | ENST00000586536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600148199.hg.1 | ENST00000590460.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600148199.hg.1 | NM_001193452.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600148199.hg.1 | NM_032349.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600148199.hg.1 | ENST00000304301.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600148199.hg.1 | ENST00000405142.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600148199.hg.1 | ENST00000586252.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600148199.hg.1 | ENST00000586536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600148199.hg.1 | ENST00000590460.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600148199.hg.1 | NM_001193452.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600148199.hg.1 | NM_032349.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600148199.hg.1 | TC1600006752.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600148200.hg.1 | ENST00000304301.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600148200.hg.1 | ENST00000405142.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600148200.hg.1 | ENST00000586252.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600148200.hg.1 | ENST00000586536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600148200.hg.1 | ENST00000590460.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600148200.hg.1 | NM_001193452.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600148200.hg.1 | NM_032349.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600148200.hg.1 | ENST00000304301.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600148200.hg.1 | ENST00000405142.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600148200.hg.1 | ENST00000586252.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600148200.hg.1 | ENST00000586536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600148200.hg.1 | ENST00000590460.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600148200.hg.1 | NM_001193452.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600148200.hg.1 | NM_032349.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600148200.hg.1 | TC1600006752.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600148201.hg.1 | ENST00000304301.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600148201.hg.1 | ENST00000405142.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600148201.hg.1 | ENST00000586252.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600148201.hg.1 | ENST00000586536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600148201.hg.1 | NM_001193452.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600148201.hg.1 | NM_032349.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600148201.hg.1 | ENST00000304301.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600148201.hg.1 | ENST00000405142.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600148201.hg.1 | ENST00000586252.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600148201.hg.1 | ENST00000586536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600148201.hg.1 | NM_001193452.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600148201.hg.1 | NM_032349.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600148201.hg.1 | TC1600006752.hg.1 | Clariom D TC |

NUDT16L1
Genomic Map