Assay Details
Gene Symbol: C4orf26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206981.1 | NP_001193910.1 | 1 - 2 | 100 | 91 | |
XM_011531668.2 | XP_011529970.1 | 2 - 3 | 1055 | 91 | ||
GenBank mRNA | AK300227.1 | - | 1 - 2 | 83 | 91 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC04000420.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR04005824.hg.1 | TC04000420.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04005824.hg.1 | ENST00000311623.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04005824.hg.1 | ENST00000435974.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04005824.hg.1 | ENST00000514064.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04005824.hg.1 | NM_001206981.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04005824.hg.1 | NM_178497.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007877.hg.1 | ENST00000311623.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007877.hg.1 | ENST00000311623.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007877.hg.1 | ENST00000311623.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007877.hg.1 | ENST00000311623.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007877.hg.1 | ENST00000311623.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007877.hg.1 | ENST00000311623.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007877.hg.1 | ENST00000311623.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007877.hg.1 | ENST00000435974.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007877.hg.1 | ENST00000435974.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007877.hg.1 | ENST00000435974.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007877.hg.1 | ENST00000435974.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007877.hg.1 | ENST00000435974.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007877.hg.1 | ENST00000435974.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007877.hg.1 | ENST00000511093.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007877.hg.1 | ENST00000511093.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007877.hg.1 | ENST00000511093.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007877.hg.1 | ENST00000511093.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007877.hg.1 | ENST00000511093.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007877.hg.1 | ENST00000511093.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007877.hg.1 | ENST00000511093.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007877.hg.1 | ENST00000514064.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007877.hg.1 | ENST00000514064.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007877.hg.1 | ENST00000514064.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007877.hg.1 | ENST00000616557.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007877.hg.1 | ENST00000616557.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007877.hg.1 | ENST00000616557.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007877.hg.1 | ENST00000616557.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007877.hg.1 | ENST00000616557.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007877.hg.1 | ENST00000616557.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007877.hg.1 | NM_001206981.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007877.hg.1 | NM_001206981.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007877.hg.1 | NM_001206981.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007877.hg.1 | NM_001206981.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007877.hg.1 | NM_001206981.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007877.hg.1 | NM_001206981.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007877.hg.1 | NM_001206981.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007877.hg.1 | NM_001206981.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007877.hg.1 | NM_001257072.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007877.hg.1 | NM_001257072.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007877.hg.1 | NM_001257072.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007877.hg.1 | NM_001257072.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007877.hg.1 | NM_001257072.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007877.hg.1 | NM_001257072.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007877.hg.1 | NM_178497.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007877.hg.1 | NM_178497.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007877.hg.1 | NM_178497.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007877.hg.1 | NM_178497.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007877.hg.1 | NM_178497.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007877.hg.1 | NM_178497.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007877.hg.1 | NM_178497.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007877.hg.1 | NR_046429.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007877.hg.1 | NR_046429.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007877.hg.1 | NR_046429.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007877.hg.1 | NR_046429.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007877.hg.1 | NR_046429.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007877.hg.1 | NR_046429.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007877.hg.1 | NR_046429.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007877.hg.1 | NR_046429.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007877.hg.1 | NR_046430.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007877.hg.1 | NR_046430.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007877.hg.1 | NR_046430.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007877.hg.1 | NR_046430.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007877.hg.1 | NR_046430.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007877.hg.1 | NR_046430.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007877.hg.1 | NR_046430.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007877.hg.1 | NR_046430.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04005825.hg.1 | TC04000420.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04005825.hg.1 | ENST00000435974.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04005825.hg.1 | ENST00000514064.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04005825.hg.1 | NM_001206981.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400154011.hg.1 | ENST00000435974.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400154011.hg.1 | ENST00000514064.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400154011.hg.1 | NM_001206981.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400154011.hg.1 | NR_046429.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400154011.hg.1 | ENST00000435974.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400154011.hg.1 | ENST00000514064.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400154011.hg.1 | NM_001206981.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400154011.hg.1 | NR_046429.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400154011.hg.1 | TC0400007877.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0400007877.hg.1 | ENST00000311623.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007877.hg.1 | ENST00000311623.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007877.hg.1 | ENST00000311623.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007877.hg.1 | ENST00000311623.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007877.hg.1 | ENST00000311623.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007877.hg.1 | ENST00000311623.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007877.hg.1 | ENST00000311623.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007877.hg.1 | ENST00000435974.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007877.hg.1 | ENST00000435974.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007877.hg.1 | ENST00000435974.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007877.hg.1 | ENST00000435974.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007877.hg.1 | ENST00000435974.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007877.hg.1 | ENST00000435974.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007877.hg.1 | ENST00000511093.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007877.hg.1 | ENST00000511093.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007877.hg.1 | ENST00000511093.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007877.hg.1 | ENST00000511093.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007877.hg.1 | ENST00000511093.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007877.hg.1 | ENST00000511093.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007877.hg.1 | ENST00000511093.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007877.hg.1 | ENST00000514064.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007877.hg.1 | ENST00000514064.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007877.hg.1 | ENST00000514064.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007877.hg.1 | ENST00000616557.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007877.hg.1 | ENST00000616557.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007877.hg.1 | ENST00000616557.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007877.hg.1 | ENST00000616557.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007877.hg.1 | ENST00000616557.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007877.hg.1 | ENST00000616557.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007877.hg.1 | NM_001206981.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007877.hg.1 | NM_001206981.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007877.hg.1 | NM_001206981.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007877.hg.1 | NM_001206981.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007877.hg.1 | NM_001206981.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007877.hg.1 | NM_001206981.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007877.hg.1 | NM_001206981.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007877.hg.1 | NM_001206981.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007877.hg.1 | NM_001257072.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007877.hg.1 | NM_001257072.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007877.hg.1 | NM_001257072.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007877.hg.1 | NM_001257072.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007877.hg.1 | NM_001257072.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007877.hg.1 | NM_001257072.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007877.hg.1 | NM_178497.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007877.hg.1 | NM_178497.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007877.hg.1 | NM_178497.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007877.hg.1 | NM_178497.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007877.hg.1 | NM_178497.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007877.hg.1 | NM_178497.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007877.hg.1 | NM_178497.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007877.hg.1 | NR_046429.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007877.hg.1 | NR_046429.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007877.hg.1 | NR_046429.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007877.hg.1 | NR_046429.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007877.hg.1 | NR_046429.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007877.hg.1 | NR_046429.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007877.hg.1 | NR_046429.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007877.hg.1 | NR_046429.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007877.hg.1 | NR_046430.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007877.hg.1 | NR_046430.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007877.hg.1 | NR_046430.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007877.hg.1 | NR_046430.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007877.hg.1 | NR_046430.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007877.hg.1 | NR_046430.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007877.hg.1 | NR_046430.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007877.hg.1 | NR_046430.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400154010.hg.1 | ENST00000311623.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400154010.hg.1 | ENST00000435974.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400154010.hg.1 | ENST00000511093.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400154010.hg.1 | ENST00000514064.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400154010.hg.1 | ENST00000616557.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400154010.hg.1 | NM_001206981.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400154010.hg.1 | NM_001257072.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400154010.hg.1 | NM_178497.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400154010.hg.1 | NR_046429.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400154010.hg.1 | NR_046430.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400154010.hg.1 | ENST00000311623.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400154010.hg.1 | ENST00000435974.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400154010.hg.1 | ENST00000511093.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400154010.hg.1 | ENST00000514064.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400154010.hg.1 | ENST00000616557.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400154010.hg.1 | NM_001206981.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400154010.hg.1 | NM_001257072.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400154010.hg.1 | NM_178497.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400154010.hg.1 | NR_046429.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400154010.hg.1 | NR_046430.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400154010.hg.1 | TC0400007877.hg.1 | Clariom D TC |
