Assay Details
| Gene Symbol: FXYD6-FXYD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243598.2 | NP_001230527.1 | 6 - 7 | 721 | 84 | |
| GenBank mRNA | CN310658.1 | - | 6 - 7 | 356 | 84 | |
More Information
Set Membership: |
|

FXYD6-FXYD2
Genomic Map