Assay Details
| Gene Symbol: IMMP2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244606.1 | NP_001231535.1 | 6 - 7 | 536 | 69 | |
| NM_032549.3 | NP_115938.1 | 5 - 6 | 850 | 69 | ||
| XM_005250630.3 | XP_005250687.1 | 7 - 8 | 618 | 69 | ||
| XM_011516605.2 | XP_011514907.1 | 7 - 8 | 634 | 69 | ||
| XM_017012705.1 | XP_016868194.1 | 6 - 7 | 588 | 69 | ||
| GenBank mRNA | AF359563.1 | - | 5 - 6 | 850 | 69 | |
| AK311968.1 | - | 4 - 5 | 501 | 69 | ||
More Information
Set Membership: |
|

IMMP2L
Genomic Map