Assay Details
| Gene Symbol: KRTDAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244847.1 | NP_001231776.1 | 1 - 2 | 180 | 69 | |
| NM_207392.2 | NP_997275.1 | 1 - 2 | 180 | 69 | ||
| GenBank mRNA | AJ293408.1 | - | 1 - 2 | 141 | 69 | |
| AY359007.1 | - | 1 - 2 | 103 | 69 | ||
| BC130501.1 | - | 1 - 2 | 180 | 40146532 | 69 | |
| W95920.1 | - | 1 - 2 | 68 | |||
More Information
Set Membership: |
|

KRTDAP
Genomic Map