Assay Details
| Gene Symbol: PCMT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252049.1 | NP_001238978.1 | 4 - 5 | 500 | 75 | |
| NM_001252051.1 | NP_001238980.1 | 3 - 4 | 395 | 75 | ||
| NM_001252053.1 | NP_001238982.1 | 4 - 5 | 500 | 75 | ||
| NM_005389.2 | NP_005380.2 | 4 - 5 | 500 | 75 | ||
| XM_011535868.2 | XP_011534170.1 | 4 - 5 | 701 | 75 | ||
| GenBank mRNA | AK129778.1 | - | 4 - 5 | 416 | 75 | |
| AK289724.1 | - | 4 - 5 | 468 | 75 | ||
| BC007501.2 | - | 4 - 5 | 442 | 3010742 | 75 | |
| BC008748.2 | - | 4 - 5 | 378 | 3346722 | 75 | |
| CX786396.1 | - | 4 - 5 | 386 | 75 | ||
| D13892.1 | - | 4 - 5 | 443 | 75 | ||
| D25545.1 | - | 4 - 5 | 366 | 75 | ||
| D25546.1 | - | 4 - 5 | 366 | 75 | ||
| D25547.1 | - | 4 - 5 | 366 | 75 | ||
| DC339361.1 | - | 4 - 5 | 496 | 75 | ||
| DC356391.1 | - | 4 - 5 | 483 | 75 | ||
| M93008.1 | - | 4 - 5 | 330 | 75 | ||
More Information
Set Membership: |
|

PCMT1
Genomic Map