Assay Details
| Gene Symbol: ADAM7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003817.3 | NP_003808.2 | 7 - 8 | 753 | 107 | |
| XM_017013943.1 | XP_016869432.1 | 7 - 8 | 640 | 107 | ||
| GenBank mRNA | AF090327.1 | - | 2 - 3 | 142 | 107 | |
| AF215824.1 | - | 7 - 8 | 724 | 107 | ||
| AK292492.1 | - | 7 - 8 | 757 | 107 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08000192.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR08002573.hg.1 | TC08000192.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08002573.hg.1 | ENST00000175238.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08002573.hg.1 | ENST00000335595.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08002573.hg.1 | ENST00000380789.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08002573.hg.1 | ENST00000520720.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08002573.hg.1 | NM_003817.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000175238.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000175238.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000175238.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000175238.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000175238.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000175238.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000175238.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000175238.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000175238.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000175238.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000175238.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000175238.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000175238.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000175238.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000175238.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000175238.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000175238.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000175238.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000175238.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000175238.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000175238.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000175238.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000175238.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000175238.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000380789.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000380789.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000380789.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000380789.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000380789.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000380789.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000380789.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000380789.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000380789.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000380789.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000380789.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000380789.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000380789.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000380789.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000380789.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000380789.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000380789.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000380789.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000380789.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000380789.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000380789.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000380789.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000380789.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000380789.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000441335.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000441335.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000441335.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000441335.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000441335.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000441335.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000441335.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000441335.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000441335.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000441335.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000441335.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000520720.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000520720.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000520720.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000520720.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000520720.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000520720.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000520720.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000520720.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000520720.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000520720.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000520720.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000520720.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000520720.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000520720.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | ENST00000520720.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | NM_003817.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | NM_003817.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | NM_003817.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | NM_003817.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | NM_003817.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | NM_003817.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | NM_003817.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | NM_003817.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | NM_003817.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | NM_003817.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | NM_003817.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | NM_003817.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | NM_003817.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | NM_003817.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | NM_003817.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | NM_003817.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | NM_003817.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | NM_003817.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | NM_003817.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | NM_003817.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | NM_003817.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | NM_003817.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | NM_003817.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | NM_003817.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007043.hg.1 | NM_003817.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000175238.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000175238.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000175238.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000175238.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000175238.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000175238.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000175238.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000175238.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000175238.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000175238.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000175238.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000175238.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000175238.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000175238.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000175238.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000175238.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000175238.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000175238.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000175238.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000175238.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000175238.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000175238.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000175238.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000175238.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000380789.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000380789.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000380789.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000380789.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000380789.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000380789.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000380789.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000380789.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000380789.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000380789.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000380789.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000380789.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000380789.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000380789.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000380789.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000380789.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000380789.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000380789.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000380789.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000380789.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000380789.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000380789.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000380789.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000380789.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000441335.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000441335.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000441335.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000441335.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000441335.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000441335.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000441335.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000441335.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000441335.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000441335.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000441335.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000520720.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000520720.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000520720.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000520720.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000520720.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000520720.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000520720.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000520720.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000520720.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000520720.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000520720.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000520720.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000520720.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000520720.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | ENST00000520720.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | NM_003817.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | NM_003817.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | NM_003817.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | NM_003817.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | NM_003817.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | NM_003817.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | NM_003817.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | NM_003817.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | NM_003817.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | NM_003817.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | NM_003817.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | NM_003817.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | NM_003817.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | NM_003817.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | NM_003817.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | NM_003817.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | NM_003817.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | NM_003817.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | NM_003817.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | NM_003817.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | NM_003817.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | NM_003817.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | NM_003817.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | NM_003817.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007043.hg.1 | NM_003817.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800148235.hg.1 | ENST00000175238.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800148235.hg.1 | ENST00000380789.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800148235.hg.1 | ENST00000520720.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800148235.hg.1 | NM_003817.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800148235.hg.1 | ENST00000175238.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800148235.hg.1 | ENST00000380789.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800148235.hg.1 | ENST00000520720.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800148235.hg.1 | NM_003817.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800148235.hg.1 | TC0800007043.hg.1 | Clariom D TC |

ADAM7
Genomic Map