Assay Details
| Gene Symbol: EML4-ALK fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB374363.1 | - | 1690 | 113 | ||
| AB462412.1 | - | 1691 | 137 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EML4-ALK fusion| Gene Symbol: EML4-ALK fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB374363.1 | - | 1690 | 113 | ||
| AB462412.1 | - | 1691 | 137 | |||
Set Membership: |
|