Assay Details
| Gene Symbol: MIR1203 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031607.1 | 1 | 42 | 57 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | sorting nexin 11 | SNX11 | -33.7kb | Anti-sense | Chr. 17 - 48107533 - 48123064 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC105371807 | THRA1/BTR | 137.9kb | Anti-sense | Chr. 17 - 48294347 - 48307829 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC1700010984.hg.1
This assay targets the same location as HTA transcript cluster(s) TC17001638.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30925203 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30925187 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30925217 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30925224 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10347605 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10347622 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30925226 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30925204 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30925179 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10347598 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10347590 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30925185 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30925210 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10347619 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10347635 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30925205 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10347596 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10347593 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10347582 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30925207 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10347637 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30925208 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30925209 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30925220 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10347617 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10347600 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30925197 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30925183 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10347578 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30925225 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10347586 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30925222 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30925184 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10347587 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30925232 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10347613 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30925213 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30925203 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30925187 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30925232 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30925204 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10347613 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30925197 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30925210 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30925217 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30925209 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10347593 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10347600 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10347578 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10347619 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30925226 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10347586 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30925207 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10347598 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30925185 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30925179 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30925225 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30925213 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10347582 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10347617 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10347622 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10347635 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10347605 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10347590 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30925208 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30925205 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10347587 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30925224 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10347596 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10347637 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30925220 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30925183 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30925222 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30925184 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR17021910.hg.1 | ENST00000408812.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17021910.hg.1 | NR_031607.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17021910.hg.1 | TC17001638.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010984.hg.1 | ENST00000408812.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010984.hg.1 | NR_031607.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010984.hg.1 | ENST00000408812.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010984.hg.1 | NR_031607.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700188362.hg.1 | ENST00000408812.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700188362.hg.1 | NR_031607.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700188362.hg.1 | ENST00000408812.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700188362.hg.1 | NR_031607.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700188362.hg.1 | TC1700010984.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3309884 | NR_031607.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3309884 | NR_031607.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3309884 | NR_031607.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3309884 | NR_031607.1 | human_transcriptome |

MIR1203
Genomic Map