Assay Details
| Gene Symbol: LOC339666 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038918.1 | 1 - 2 | 56 | 62 | ||
| GenBank mRNA | BC041349.2 | - | 1 - 2 | 56 | 62 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RFPL3 antisense | RFPL3S |
NCRNA00005, RFPL3-AS, RFPL3-AS1, RFPL3ANT, RFPL3AS |
-5.4kb | Anti-sense | Chr. 22 - 32359906 - 32371264 on NCBI Build GRCh38 |
| Downstream | RNA 2',3'-cyclic phosphate and 5'-OH ligase | RTCB |
C22orf28, DJ149A16.6, FAAP, HSPC117 |
9.8kb | Anti-sense | Chr. 22 - 32387575 - 32412287 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22000244.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32159687 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32159694 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11477239 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32159685 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32159697 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32159704 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11477253 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11477241 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32159683 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32159680 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11477246 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32159699 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11477251 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32159684 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11477248 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32159692 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11477240 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32159681 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32159686 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11477245 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3663277 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 20670 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3663277 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3663277 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P03617133 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 1562841_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR22004612.hg.1 | ENST00000420171.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22004612.hg.1 | NR_038918.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22004612.hg.1 | TC22000244.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22004611.hg.1 | ENST00000420171.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22004611.hg.1 | ENST00000444848.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22004611.hg.1 | NR_038918.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22004611.hg.1 | TC22000244.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22004608.hg.1 | ENST00000420171.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22004608.hg.1 | NR_038918.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22004608.hg.1 | TC22000244.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22004610.hg.1 | ENST00000420171.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22004610.hg.1 | NR_038918.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22004610.hg.1 | TC22000244.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007135.hg.1 | ENST00000420171.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007135.hg.1 | ENST00000420171.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007135.hg.1 | ENST00000420171.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007135.hg.1 | ENST00000420171.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007135.hg.1 | ENST00000420171.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007135.hg.1 | ENST00000430579.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007135.hg.1 | ENST00000444848.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007135.hg.1 | ENST00000444848.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007135.hg.1 | NR_038918.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007135.hg.1 | NR_038918.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007135.hg.1 | NR_038918.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007135.hg.1 | NR_038918.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007135.hg.1 | NR_038918.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22004609.hg.1 | ENST00000420171.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22004609.hg.1 | NR_038918.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22004609.hg.1 | TC22000244.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR2200150267.hg.1 | TC2200007135.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200150267.hg.1 | ENST00000420171.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200150267.hg.1 | NR_038918.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200150267.hg.1 | ENST00000420171.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200150267.hg.1 | NR_038918.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200150269.hg.1 | TC2200007135.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200150269.hg.1 | ENST00000420171.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200150269.hg.1 | NR_038918.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200150269.hg.1 | ENST00000420171.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200150269.hg.1 | NR_038918.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200150268.hg.1 | TC2200007135.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200150268.hg.1 | ENST00000420171.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200150268.hg.1 | ENST00000444848.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200150268.hg.1 | NR_038918.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200150268.hg.1 | ENST00000420171.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200150268.hg.1 | ENST00000444848.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200150268.hg.1 | NR_038918.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200150265.hg.1 | TC2200007135.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200150265.hg.1 | ENST00000420171.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200150265.hg.1 | NR_038918.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200150265.hg.1 | ENST00000420171.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200150265.hg.1 | NR_038918.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200150266.hg.1 | TC2200007135.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200150266.hg.1 | ENST00000420171.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200150266.hg.1 | NR_038918.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200150266.hg.1 | ENST00000420171.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200150266.hg.1 | NR_038918.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007135.hg.1 | ENST00000420171.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007135.hg.1 | ENST00000420171.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007135.hg.1 | ENST00000420171.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007135.hg.1 | ENST00000420171.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007135.hg.1 | ENST00000420171.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007135.hg.1 | ENST00000430579.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007135.hg.1 | ENST00000444848.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007135.hg.1 | ENST00000444848.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007135.hg.1 | NR_038918.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007135.hg.1 | NR_038918.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007135.hg.1 | NR_038918.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007135.hg.1 | NR_038918.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007135.hg.1 | NR_038918.1 | Refseq |
| Human Genome U133 Plus 2.0 Array | 1562841_at | - | - |
| Human Genome U133 Plus 2.0 Array | 234385_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 234385_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1562841_PM_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562841_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562841_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562841_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562841_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562841_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562841_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562841_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562841_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562841_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562841_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562841_at2 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A020361 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A020361 | - | - |

LOC339666
Genomic Map