| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007249.hg.1 |
ENST00000629357.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007249.hg.1 |
ENST00000629357.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007249.hg.1 |
ENST00000629357.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007249.hg.1 |
ENST00000629357.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007249.hg.1 |
ENST00000629357.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007249.hg.1 |
ENST00000629357.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007249.hg.1 |
ENST00000629357.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007249.hg.1 |
ENST00000629357.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007249.hg.1 |
ENST00000629357.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007249.hg.1 |
ENST00000629576.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007249.hg.1 |
ENST00000629576.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007249.hg.1 |
ENST00000629576.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007249.hg.1 |
ENST00000629576.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007249.hg.1 |
NR_040085.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007249.hg.1 |
NR_040085.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007249.hg.1 |
NR_040085.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007249.hg.1 |
NR_040085.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007249.hg.1 |
NR_040085.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007249.hg.1 |
NR_040085.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007249.hg.1 |
NR_040085.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007249.hg.1 |
NR_040085.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007249.hg.1 |
NR_040085.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07003781.hg.1 |
TC07000246.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07003781.hg.1 |
ENST00000515771.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07003781.hg.1 |
NR_040085.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007249.hg.1 |
ENST00000629357.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007249.hg.1 |
ENST00000629357.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007249.hg.1 |
ENST00000629357.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007249.hg.1 |
ENST00000629357.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007249.hg.1 |
ENST00000629357.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007249.hg.1 |
ENST00000629357.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007249.hg.1 |
ENST00000629357.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007249.hg.1 |
ENST00000629357.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007249.hg.1 |
ENST00000629357.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007249.hg.1 |
ENST00000629576.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007249.hg.1 |
ENST00000629576.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007249.hg.1 |
ENST00000629576.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007249.hg.1 |
ENST00000629576.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007249.hg.1 |
NR_040085.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007249.hg.1 |
NR_040085.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007249.hg.1 |
NR_040085.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007249.hg.1 |
NR_040085.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007249.hg.1 |
NR_040085.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007249.hg.1 |
NR_040085.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007249.hg.1 |
NR_040085.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007249.hg.1 |
NR_040085.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007249.hg.1 |
NR_040085.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700150343.hg.1 |
ENST00000629357.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700150343.hg.1 |
NR_040085.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700150343.hg.1 |
ENST00000629357.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700150343.hg.1 |
NR_040085.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700150343.hg.1 |
TC0700007249.hg.1 |
Clariom D TC |