Assay Details
| Gene Symbol: TRG-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040085.1 | 3 - 4 | 438 | 68 | ||
| GenBank mRNA | AK125646.1 | - | 3 - 4 | 400 | 68 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | T cell receptor gamma variable 5 | TRGV5 |
TCRGV5, V1S5 |
-522.0 | Anti-sense | Chr. 7 - 38349355 - 38349824 on NCBI Build GRCh38 |
| Downstream | amphiphysin | AMPH | AMPH1 | 5.1kb | Anti-sense | Chr. 7 - 38383694 - 38631567 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07000246.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25649852 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05428833 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25650030 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05428837 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25649836 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05428836 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25649845 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25650031 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25649835 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05428828 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05428813 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15931461 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25649837 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15931286 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05428811 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25650026 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05428835 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25649847 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25650033 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05428807 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25649859 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 137352 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 76010 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 184209 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 52890 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 722381 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 800223 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 477438 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 412087 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 845661 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 341289 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 136211 | AK226138.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 136211 | AK290046.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 136211 | AY366498.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 136211 | BC117116.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 136211 | D17516.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 136211 | NM_001118.3 | refseq_mrna_human_NM |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 166496 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 62779 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 286109 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 26552 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 166444 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 65560 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 62507 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P17913808 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P17913790 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P17913808 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 58978 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P17913801 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P17913808 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P17913797 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P17913790 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P37994816 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P01695573 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P17913808 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P17913790 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR07003782.hg.1 | TC07000246.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07003782.hg.1 | ENST00000515771.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07003782.hg.1 | NR_040085.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007249.hg.1 | ENST00000629357.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007249.hg.1 | ENST00000629357.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007249.hg.1 | ENST00000629357.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007249.hg.1 | ENST00000629357.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007249.hg.1 | ENST00000629357.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007249.hg.1 | ENST00000629357.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007249.hg.1 | ENST00000629357.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007249.hg.1 | ENST00000629357.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007249.hg.1 | ENST00000629357.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007249.hg.1 | ENST00000629576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007249.hg.1 | ENST00000629576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007249.hg.1 | ENST00000629576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007249.hg.1 | ENST00000629576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007249.hg.1 | NR_040085.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007249.hg.1 | NR_040085.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007249.hg.1 | NR_040085.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007249.hg.1 | NR_040085.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007249.hg.1 | NR_040085.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007249.hg.1 | NR_040085.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007249.hg.1 | NR_040085.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007249.hg.1 | NR_040085.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007249.hg.1 | NR_040085.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700150348.hg.1 | ENST00000629357.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700150348.hg.1 | NR_040085.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700150348.hg.1 | ENST00000629357.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700150348.hg.1 | NR_040085.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700150348.hg.1 | TC0700007249.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700150345.hg.1 | ENST00000629357.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700150345.hg.1 | NR_040085.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700150345.hg.1 | ENST00000629357.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700150345.hg.1 | NR_040085.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700150345.hg.1 | TC0700007249.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700150344.hg.1 | ENST00000629357.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700150344.hg.1 | NR_040085.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700150344.hg.1 | ENST00000629357.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700150344.hg.1 | NR_040085.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700150344.hg.1 | TC0700007249.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700007249.hg.1 | ENST00000629357.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007249.hg.1 | ENST00000629357.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007249.hg.1 | ENST00000629357.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007249.hg.1 | ENST00000629357.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007249.hg.1 | ENST00000629357.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007249.hg.1 | ENST00000629357.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007249.hg.1 | ENST00000629357.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007249.hg.1 | ENST00000629357.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007249.hg.1 | ENST00000629357.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007249.hg.1 | ENST00000629576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007249.hg.1 | ENST00000629576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007249.hg.1 | ENST00000629576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007249.hg.1 | ENST00000629576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007249.hg.1 | NR_040085.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007249.hg.1 | NR_040085.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007249.hg.1 | NR_040085.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007249.hg.1 | NR_040085.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007249.hg.1 | NR_040085.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007249.hg.1 | NR_040085.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007249.hg.1 | NR_040085.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007249.hg.1 | NR_040085.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007249.hg.1 | NR_040085.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700150347.hg.1 | ENST00000629357.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700150347.hg.1 | NR_040085.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700150347.hg.1 | ENST00000629357.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700150347.hg.1 | NR_040085.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700150347.hg.1 | TC0700007249.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700150346.hg.1 | ENST00000629357.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700150346.hg.1 | NR_040085.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700150346.hg.1 | ENST00000629357.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700150346.hg.1 | NR_040085.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700150346.hg.1 | TC0700007249.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A034222 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A034222 | - | - |

TRG-AS1
Genomic Map