Assay Details
Gene Symbol: RAET1E-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045127.1 | 1 - 2 | 63 | 59 | ||
GenBank mRNA | FY211027.1 | - | 1 - 2 | 41 | 59 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC06001086.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR06012849.hg.1 | TC06001086.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06012849.hg.1 | ENST00000446954.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06012849.hg.1 | NR_045127.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06012848.hg.1 | TC06001086.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06012848.hg.1 | ENST00000446954.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06012848.hg.1 | NR_045127.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009813.hg.1 | ENST00000446954.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009813.hg.1 | ENST00000446954.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009813.hg.1 | ENST00000446954.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009813.hg.1 | ENST00000446954.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009813.hg.1 | ENST00000596229.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009813.hg.1 | ENST00000596229.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009813.hg.1 | ENST00000596229.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009813.hg.1 | ENST00000596229.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009813.hg.1 | ENST00000596229.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009813.hg.1 | ENST00000605899.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009813.hg.1 | ENST00000605899.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009813.hg.1 | ENST00000605899.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009813.hg.1 | ENST00000605899.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009813.hg.1 | ENST00000606915.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009813.hg.1 | ENST00000606915.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009813.hg.1 | ENST00000606915.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009813.hg.1 | ENST00000606915.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009813.hg.1 | ENST00000606915.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009813.hg.1 | ENST00000606915.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009813.hg.1 | ENST00000606915.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009813.hg.1 | NR_045126.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009813.hg.1 | NR_045126.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009813.hg.1 | NR_045126.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009813.hg.1 | NR_045126.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009813.hg.1 | NR_045126.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009813.hg.1 | NR_045126.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009813.hg.1 | NR_045126.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009813.hg.1 | NR_045127.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009813.hg.1 | NR_045127.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009813.hg.1 | NR_045127.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009813.hg.1 | NR_045127.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009813.hg.1 | ENST00000446954.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009813.hg.1 | ENST00000446954.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009813.hg.1 | ENST00000446954.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009813.hg.1 | ENST00000446954.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009813.hg.1 | ENST00000596229.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009813.hg.1 | ENST00000596229.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009813.hg.1 | ENST00000596229.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009813.hg.1 | ENST00000596229.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009813.hg.1 | ENST00000596229.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009813.hg.1 | ENST00000605899.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009813.hg.1 | ENST00000605899.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009813.hg.1 | ENST00000605899.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009813.hg.1 | ENST00000605899.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009813.hg.1 | ENST00000606915.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009813.hg.1 | ENST00000606915.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009813.hg.1 | ENST00000606915.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009813.hg.1 | ENST00000606915.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009813.hg.1 | ENST00000606915.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009813.hg.1 | ENST00000606915.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009813.hg.1 | ENST00000606915.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009813.hg.1 | NR_045126.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009813.hg.1 | NR_045126.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009813.hg.1 | NR_045126.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009813.hg.1 | NR_045126.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009813.hg.1 | NR_045126.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009813.hg.1 | NR_045126.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009813.hg.1 | NR_045126.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009813.hg.1 | NR_045127.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009813.hg.1 | NR_045127.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009813.hg.1 | NR_045127.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009813.hg.1 | NR_045127.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600167192.hg.1 | ENST00000446954.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600167192.hg.1 | NR_045127.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600167192.hg.1 | ENST00000446954.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600167192.hg.1 | NR_045127.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600167192.hg.1 | TC0600009813.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600167193.hg.1 | ENST00000446954.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600167193.hg.1 | NR_045127.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600167193.hg.1 | ENST00000446954.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600167193.hg.1 | NR_045127.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600167193.hg.1 | TC0600009813.hg.1 | Clariom D TC |
