Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009439.hg.1 |
ENST00000461291.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009439.hg.1 |
ENST00000461291.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009439.hg.1 |
ENST00000461291.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009439.hg.1 |
NR_046231.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009439.hg.1 |
NR_046231.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009439.hg.1 |
NR_046231.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009439.hg.1 |
NR_046231.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009439.hg.1 |
ENST00000461291.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009439.hg.1 |
ENST00000461291.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009439.hg.1 |
ENST00000461291.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009439.hg.1 |
NR_046231.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009439.hg.1 |
NR_046231.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009439.hg.1 |
NR_046231.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009439.hg.1 |
NR_046231.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000164494.hg.1 |
ENST00000461291.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000164494.hg.1 |
NR_046231.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000164494.hg.1 |
ENST00000461291.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000164494.hg.1 |
NR_046231.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000164494.hg.1 |
TC1000009439.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000164496.hg.1 |
ENST00000461291.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000164496.hg.1 |
NR_046231.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000164496.hg.1 |
ENST00000461291.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000164496.hg.1 |
NR_046231.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000164496.hg.1 |
TC1000009439.hg.1 |
Clariom D TC |