Assay Details
Gene Symbol: LINC01168 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_046231.1 | 2 - 3 | 4133 | 54 | ||
GenBank mRNA | AK125849.1 | - | 2 - 3 | 4133 | 54 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | long intergenic non-protein coding RNA 1166 | LINC01166 | -5.8kb | Anti-sense | Chr. 10 - 132943967 - 132965289 on NCBI Build GRCh38 | |
Downstream | adhesion G protein-coupled receptor A1 | ADGRA1 | GPR123 | 94.4kb | Sense | Chr. 10 - 133070929 - 133131675 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07591402 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07591375 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07591430 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27983092 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16402485 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27983124 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07591425 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07591456 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07591429 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07591414 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07591377 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07591444 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07591386 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16402492 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16402489 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16402479 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27983156 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07591432 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07591412 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07591451 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16402501 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07591379 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07591411 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07591455 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07591416 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07591431 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07591392 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07591384 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27983149 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07591427 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07591457 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07591406 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07591395 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16402500 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27983144 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27983126 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27983073 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07591388 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07591435 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27983080 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07591438 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27983114 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07591380 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07591385 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07591420 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27983142 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07591419 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07591448 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07591460 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27983118 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27983129 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27983145 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07591423 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16402474 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27983147 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27983066 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07591394 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27983103 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07591445 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27983075 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07591391 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27983098 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07591401 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07591408 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27983111 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16402493 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27983158 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07591415 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07591454 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07591453 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07591458 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07591390 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27983140 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07591404 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07591397 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3283974 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 6601 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P02377608 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3283974 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3283974 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P02377608 | - | - |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 51697 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P02377614 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P02377608 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P18661572 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P32806318 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1000009439.hg.1 | ENST00000461291.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009439.hg.1 | ENST00000461291.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009439.hg.1 | ENST00000461291.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009439.hg.1 | NR_046231.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009439.hg.1 | NR_046231.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009439.hg.1 | NR_046231.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009439.hg.1 | NR_046231.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000164497.hg.1 | ENST00000461291.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000164497.hg.1 | NR_046231.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000164497.hg.1 | ENST00000461291.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000164497.hg.1 | NR_046231.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000164497.hg.1 | TC1000009439.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1000164498.hg.1 | NR_046231.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000164498.hg.1 | NR_046231.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000164498.hg.1 | TC1000009439.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1000009439.hg.1 | ENST00000461291.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009439.hg.1 | ENST00000461291.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009439.hg.1 | ENST00000461291.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009439.hg.1 | NR_046231.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009439.hg.1 | NR_046231.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009439.hg.1 | NR_046231.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009439.hg.1 | NR_046231.1 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1661288 | AK125849.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1661288 | AK125849.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1661288 | AK125849.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1661288 | AK125849.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1661288 | AK125849.1 | human_transcriptome |
