Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008811.hg.1 |
ENST00000412178.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008811.hg.1 |
ENST00000412178.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008811.hg.1 |
ENST00000412178.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008811.hg.1 |
NR_046853.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008811.hg.1 |
NR_046853.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008811.hg.1 |
NR_046853.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000161530.hg.1 |
ENST00000412178.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000161530.hg.1 |
NR_046853.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000161530.hg.1 |
TC2000008811.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000161530.hg.1 |
ENST00000412178.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000161530.hg.1 |
NR_046853.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008811.hg.1 |
ENST00000412178.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008811.hg.1 |
ENST00000412178.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008811.hg.1 |
ENST00000412178.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008811.hg.1 |
NR_046853.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008811.hg.1 |
NR_046853.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008811.hg.1 |
NR_046853.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000161531.hg.1 |
ENST00000412178.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000161531.hg.1 |
NR_046853.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000161531.hg.1 |
TC2000008811.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000161531.hg.1 |
ENST00000412178.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000161531.hg.1 |
NR_046853.2 |
Refseq |