Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007639.hg.1 |
ENST00000446314.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007639.hg.1 |
ENST00000446314.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007639.hg.1 |
ENST00000446314.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007639.hg.1 |
NR_046871.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007639.hg.1 |
NR_046871.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007639.hg.1 |
NR_046871.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007639.hg.1 |
NR_046871.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007639.hg.1 |
NR_046871.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007639.hg.1 |
NR_046871.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007639.hg.1 |
NR_046871.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007639.hg.1 |
ENST00000446314.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007639.hg.1 |
ENST00000446314.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007639.hg.1 |
ENST00000446314.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007639.hg.1 |
NR_046871.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007639.hg.1 |
NR_046871.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007639.hg.1 |
NR_046871.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007639.hg.1 |
NR_046871.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007639.hg.1 |
NR_046871.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007639.hg.1 |
NR_046871.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007639.hg.1 |
NR_046871.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300151585.hg.1 |
NR_046871.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300151585.hg.1 |
NR_046871.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300151585.hg.1 |
TC1300007639.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300151584.hg.1 |
NR_046871.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300151584.hg.1 |
NR_046871.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300151584.hg.1 |
TC1300007639.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300151586.hg.1 |
NR_046871.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300151586.hg.1 |
NR_046871.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300151586.hg.1 |
TC1300007639.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300151589.hg.1 |
ENST00000446314.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300151589.hg.1 |
NR_046871.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300151589.hg.1 |
ENST00000446314.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300151589.hg.1 |
NR_046871.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300151589.hg.1 |
TC1300007639.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300151588.hg.1 |
ENST00000446314.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300151588.hg.1 |
NR_046871.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300151588.hg.1 |
ENST00000446314.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300151588.hg.1 |
NR_046871.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300151588.hg.1 |
TC1300007639.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300151587.hg.1 |
ENST00000446314.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300151587.hg.1 |
NR_046871.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300151587.hg.1 |
ENST00000446314.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300151587.hg.1 |
NR_046871.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300151587.hg.1 |
TC1300007639.hg.1 |
Clariom D TC |