Assay Details
Gene Symbol: LINC00333 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_046871.1 | 6 - 7 | 747 | 76 | ||
GenBank mRNA | BI821261.1 | - | 6 - 7 | 746 | 76 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | SLIT and NTRK like family member 1 | SLITRK1 |
LRRC12, TTM |
-680.4kb | Anti-sense | Chr. 13 - 83877205 - 83882393 on NCBI Build GRCh38 |
Downstream | long intergenic non-protein coding RNA 375 | LINC00375 | 458.3kb | Anti-sense | Chr. 13 - 85065087 - 85148057 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
