Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007648.hg.1 |
ENST00000424926.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007648.hg.1 |
ENST00000424926.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007648.hg.1 |
ENST00000424926.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007648.hg.1 |
ENST00000424926.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007648.hg.1 |
ENST00000424926.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007648.hg.1 |
ENST00000424926.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007648.hg.1 |
NR_046989.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007648.hg.1 |
NR_046989.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007648.hg.1 |
NR_046989.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007648.hg.1 |
NR_046989.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007648.hg.1 |
NR_046989.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007648.hg.1 |
NR_046989.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007648.hg.1 |
NR_046989.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007648.hg.1 |
ENST00000424926.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007648.hg.1 |
ENST00000424926.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007648.hg.1 |
ENST00000424926.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007648.hg.1 |
ENST00000424926.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007648.hg.1 |
ENST00000424926.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007648.hg.1 |
ENST00000424926.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007648.hg.1 |
NR_046989.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007648.hg.1 |
NR_046989.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007648.hg.1 |
NR_046989.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007648.hg.1 |
NR_046989.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007648.hg.1 |
NR_046989.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007648.hg.1 |
NR_046989.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007648.hg.1 |
NR_046989.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300151618.hg.1 |
ENST00000424926.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300151618.hg.1 |
NR_046989.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300151618.hg.1 |
ENST00000424926.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300151618.hg.1 |
NR_046989.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300151618.hg.1 |
TC1300007648.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300151623.hg.1 |
ENST00000424926.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300151623.hg.1 |
NR_046989.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300151623.hg.1 |
ENST00000424926.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300151623.hg.1 |
NR_046989.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300151623.hg.1 |
TC1300007648.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300151625.hg.1 |
ENST00000424926.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300151625.hg.1 |
NR_046989.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300151625.hg.1 |
ENST00000424926.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300151625.hg.1 |
NR_046989.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300151625.hg.1 |
TC1300007648.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300151619.hg.1 |
ENST00000424926.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300151619.hg.1 |
NR_046989.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300151619.hg.1 |
ENST00000424926.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300151619.hg.1 |
NR_046989.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300151619.hg.1 |
TC1300007648.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300151624.hg.1 |
ENST00000424926.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300151624.hg.1 |
NR_046989.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300151624.hg.1 |
ENST00000424926.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300151624.hg.1 |
NR_046989.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300151624.hg.1 |
TC1300007648.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300151627.hg.1 |
NR_046989.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300151627.hg.1 |
NR_046989.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300151627.hg.1 |
TC1300007648.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300151626.hg.1 |
ENST00000424926.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300151626.hg.1 |
NR_046989.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300151626.hg.1 |
ENST00000424926.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300151626.hg.1 |
NR_046989.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300151626.hg.1 |
TC1300007648.hg.1 |
Clariom D TC |