| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009434.hg.1 |
ENST00000435401.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009434.hg.1 |
ENST00000435401.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009434.hg.1 |
ENST00000435401.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009434.hg.1 |
ENST00000435401.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009434.hg.1 |
ENST00000435401.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009434.hg.1 |
NR_046991.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009434.hg.1 |
NR_046991.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009434.hg.1 |
NR_046991.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009434.hg.1 |
NR_046991.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009434.hg.1 |
NR_046991.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007689.hg.1 |
ENST00000434117.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007689.hg.1 |
ENST00000434117.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007689.hg.1 |
ENST00000434117.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007689.hg.1 |
ENST00000434117.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007689.hg.1 |
ENST00000434117.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007689.hg.1 |
ENST00000434117.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300163195.hg.1 |
ENST00000435401.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300163195.hg.1 |
NR_046991.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300163195.hg.1 |
ENST00000435401.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300163195.hg.1 |
NR_046991.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300163195.hg.1 |
TC1300009434.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1300163192.hg.1 |
ENST00000435401.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300163192.hg.1 |
NR_046991.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300163192.hg.1 |
ENST00000435401.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300163192.hg.1 |
NR_046991.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300163192.hg.1 |
TC1300009434.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1300163193.hg.1 |
ENST00000435401.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300163193.hg.1 |
NR_046991.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300163193.hg.1 |
ENST00000435401.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300163193.hg.1 |
NR_046991.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300163193.hg.1 |
TC1300009434.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1300163198.hg.1 |
ENST00000435401.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300163198.hg.1 |
NR_046991.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300163198.hg.1 |
ENST00000435401.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300163198.hg.1 |
NR_046991.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300163198.hg.1 |
TC1300009434.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1300009434.hg.1 |
ENST00000435401.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009434.hg.1 |
ENST00000435401.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009434.hg.1 |
ENST00000435401.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009434.hg.1 |
ENST00000435401.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009434.hg.1 |
ENST00000435401.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009434.hg.1 |
NR_046991.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009434.hg.1 |
NR_046991.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009434.hg.1 |
NR_046991.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009434.hg.1 |
NR_046991.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009434.hg.1 |
NR_046991.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300163200.hg.1 |
NR_046991.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300163200.hg.1 |
NR_046991.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300163200.hg.1 |
TC1300009434.hg.1 |
Clariom D TC |