Assay Details
| Gene Symbol: JRKL-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_047481.1 | 4 - 5 | 551 | 60 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | JRK-like | JRKL | HHMJG | -49.7kb | Anti-sense | Chr. 11 - 96389994 - 96393563 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC105369443 | LOC105369443 | 60.2kb | Anti-sense | Chr. 11 - 96508425 - 96514750 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|

JRKL-AS1
Genomic Map