Assay Details
| Gene Symbol: LOC729732 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_047662.2 | 8 - 9 | 1999 | 80 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC100506990 | LOC100506990 | Anti-sense | Chr. 8 - 12437013 - 12566845 on NCBI Build GRCh38 | ||
| Downstream | microRNA 5692a-2 | MIR5692A2 | 138.1kb | Anti-sense | Chr. 8 - 12719132 - 12719190 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3226395 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3211793 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3243500 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3243502 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3226407 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200012751.hg.1 | ENST00000420040.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012751.hg.1 | ENST00000420040.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012751.hg.1 | ENST00000420040.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012751.hg.1 | ENST00000420040.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012751.hg.1 | ENST00000420040.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012751.hg.1 | ENST00000420040.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012751.hg.1 | ENST00000536539.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012751.hg.1 | ENST00000536539.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012751.hg.1 | ENST00000536539.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012751.hg.1 | ENST00000536560.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012751.hg.1 | ENST00000536560.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012751.hg.1 | ENST00000537659.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012751.hg.1 | ENST00000537659.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012751.hg.1 | ENST00000537659.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012751.hg.1 | ENST00000537659.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012751.hg.1 | ENST00000537659.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012751.hg.1 | ENST00000537659.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012751.hg.1 | ENST00000537659.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012751.hg.1 | ENST00000537659.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012751.hg.1 | ENST00000537659.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012751.hg.1 | ENST00000537659.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012751.hg.1 | ENST00000537659.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012751.hg.1 | ENST00000537659.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012751.hg.1 | ENST00000538304.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012751.hg.1 | ENST00000538304.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012751.hg.1 | ENST00000538304.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012751.hg.1 | ENST00000538304.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012751.hg.1 | ENST00000538304.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012751.hg.1 | ENST00000538304.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012751.hg.1 | ENST00000538304.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012751.hg.1 | ENST00000538304.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012751.hg.1 | ENST00000538304.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012751.hg.1 | ENST00000538304.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012751.hg.1 | ENST00000539348.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012751.hg.1 | ENST00000539348.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012751.hg.1 | ENST00000539348.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012751.hg.1 | ENST00000539348.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012751.hg.1 | ENST00000539348.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012751.hg.1 | ENST00000539348.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012751.hg.1 | ENST00000539348.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012751.hg.1 | ENST00000539348.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012751.hg.1 | ENST00000539348.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012751.hg.1 | ENST00000539348.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012751.hg.1 | NR_024420.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012751.hg.1 | NR_024420.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012751.hg.1 | NR_024420.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012751.hg.1 | NR_024420.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012751.hg.1 | NR_024420.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012751.hg.1 | NR_024420.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800165637.hg.1 | NR_047662.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800165637.hg.1 | NR_047662.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800165637.hg.1 | TC0800009661.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200012751.hg.1 | ENST00000420040.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012751.hg.1 | ENST00000420040.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012751.hg.1 | ENST00000420040.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012751.hg.1 | ENST00000420040.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012751.hg.1 | ENST00000420040.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012751.hg.1 | ENST00000420040.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012751.hg.1 | ENST00000536539.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012751.hg.1 | ENST00000536539.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012751.hg.1 | ENST00000536539.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012751.hg.1 | ENST00000536560.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012751.hg.1 | ENST00000536560.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012751.hg.1 | ENST00000537659.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012751.hg.1 | ENST00000537659.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012751.hg.1 | ENST00000537659.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012751.hg.1 | ENST00000537659.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012751.hg.1 | ENST00000537659.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012751.hg.1 | ENST00000537659.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012751.hg.1 | ENST00000537659.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012751.hg.1 | ENST00000537659.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012751.hg.1 | ENST00000537659.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012751.hg.1 | ENST00000537659.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012751.hg.1 | ENST00000537659.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012751.hg.1 | ENST00000537659.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012751.hg.1 | ENST00000538304.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012751.hg.1 | ENST00000538304.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012751.hg.1 | ENST00000538304.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012751.hg.1 | ENST00000538304.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012751.hg.1 | ENST00000538304.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012751.hg.1 | ENST00000538304.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012751.hg.1 | ENST00000538304.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012751.hg.1 | ENST00000538304.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012751.hg.1 | ENST00000538304.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012751.hg.1 | ENST00000538304.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012751.hg.1 | ENST00000539348.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012751.hg.1 | ENST00000539348.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012751.hg.1 | ENST00000539348.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012751.hg.1 | ENST00000539348.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012751.hg.1 | ENST00000539348.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012751.hg.1 | ENST00000539348.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012751.hg.1 | ENST00000539348.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012751.hg.1 | ENST00000539348.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012751.hg.1 | ENST00000539348.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012751.hg.1 | ENST00000539348.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012751.hg.1 | NR_024420.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012751.hg.1 | NR_024420.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012751.hg.1 | NR_024420.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012751.hg.1 | NR_024420.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012751.hg.1 | NR_024420.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012751.hg.1 | NR_024420.1 | Refseq |
| Human Genome U95 Set | 80360_at | - | - |
| Human Genome U95 Set | 80360_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1791297 | BC037255.2 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1791297 | AK124896.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1791297 | BC037255.2 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1791297 | AK124896.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1791297 | BC037255.2 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1791297 | AK124896.1 | human_transcriptome |

LOC729732
Genomic Map