Assay Details
| Gene Symbol: MS4A14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079692.2 | NP_001073160.1 | 2 - 3 | 835 | 92 | |
| NM_001261827.1 | NP_001248756.1 | 2 - 3 | 835 | 92 | ||
| NR_049731.1 | 2 - 3 | 835 | 92 | |||
| NR_049733.1 | 2 - 3 | 835 | 92 | |||
| GenBank mRNA | AK057418.1 | - | 2 - 3 | 381 | 92 | |
| AY094611.1 | - | 2 - 3 | 419 | 92 | ||
| AY584610.1 | - | 2 - 3 | 419 | 92 | ||
| DB082055.1 | - | 2 - 3 | 385 | 92 | ||
More Information
Set Membership: |
|

MS4A14
Genomic Map