Assay Details
| Gene Symbol: SNX17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_049787.1 | 1 - 2 | 315 | 91 | ||
| NR_049788.1 | 1 - 2 | 315 | 91 | |||
| GenBank mRNA | AK295278.1 | - | 1 - 2 | 194 | 91 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | eukaryotic translation initiation factor 2B subunit delta | EIF2B4 |
EIF-2B, EIF2B, EIF2Bdelta |
-39.0 | Anti-sense | Chr. 2 - 27364352 - 27370457 on NCBI Build GRCh38 |
| Downstream | zinc finger protein 513 | ZNF513 |
HMFT0656, RP58 |
3.9kb | Anti-sense | Chr. 2 - 27377231 - 27380866 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK295278.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
cardiac septum development
cholesterol catabolic process
intracellular protein transport
receptor-mediated endocytosis
signal transduction
endosomal transport
regulation of endocytosis
aorta development
coronary vasculature development
retrograde transport, endosome to plasma membrane
receptor binding
protein binding
protein C-terminus binding
phosphatidylinositol binding
low-density lipoprotein particle receptor binding
cardiac septum development
cholesterol catabolic process
intracellular protein transport
receptor-mediated endocytosis
signal transduction
endosomal transport
regulation of endocytosis
aorta development
coronary vasculature development
retrograde transport, endosome to plasma membrane
receptor binding
protein binding
protein C-terminus binding
phosphatidylinositol binding
low-density lipoprotein particle receptor binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Clariom D Assay | PSR0200148417.hg.1 | ENST00000537606.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200148417.hg.1 | NM_001267059.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148417.hg.1 | NM_001267060.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148417.hg.1 | NM_001267061.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148417.hg.1 | NM_014748.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148417.hg.1 | NR_049782.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148417.hg.1 | NR_049783.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148417.hg.1 | NR_049784.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148417.hg.1 | NR_049785.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148417.hg.1 | NR_049786.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148417.hg.1 | NR_049787.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148417.hg.1 | NR_049788.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148417.hg.1 | ENST00000537606.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200148417.hg.1 | NM_001267059.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148417.hg.1 | NM_001267060.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148417.hg.1 | NM_001267061.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148417.hg.1 | NM_014748.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148417.hg.1 | NR_049782.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148417.hg.1 | NR_049783.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148417.hg.1 | NR_049784.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148417.hg.1 | NR_049785.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148417.hg.1 | NR_049786.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148417.hg.1 | NR_049787.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148417.hg.1 | NR_049788.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148417.hg.1 | TC0200007064.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200148429.hg.1 | ENST00000233575.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200148429.hg.1 | ENST00000427123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200148429.hg.1 | ENST00000440760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200148429.hg.1 | ENST00000453453.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200148429.hg.1 | ENST00000464279.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200148429.hg.1 | ENST00000484886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200148429.hg.1 | ENST00000489402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200148429.hg.1 | ENST00000494893.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200148429.hg.1 | ENST00000537606.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200148429.hg.1 | NM_001267059.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148429.hg.1 | NM_001267060.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148429.hg.1 | NM_014748.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148429.hg.1 | NR_049782.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148429.hg.1 | NR_049783.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148429.hg.1 | NR_049784.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148429.hg.1 | NR_049785.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148429.hg.1 | NR_049786.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148429.hg.1 | NR_049787.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148429.hg.1 | NR_049788.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148429.hg.1 | ENST00000233575.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200148429.hg.1 | ENST00000427123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200148429.hg.1 | ENST00000440760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200148429.hg.1 | ENST00000453453.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200148429.hg.1 | ENST00000464279.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200148429.hg.1 | ENST00000484886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200148429.hg.1 | ENST00000489402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200148429.hg.1 | ENST00000494893.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200148429.hg.1 | ENST00000537606.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200148429.hg.1 | NM_001267059.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148429.hg.1 | NM_001267060.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148429.hg.1 | NM_014748.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148429.hg.1 | NR_049782.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148429.hg.1 | NR_049783.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148429.hg.1 | NR_049784.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148429.hg.1 | NR_049785.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148429.hg.1 | NR_049786.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148429.hg.1 | NR_049787.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148429.hg.1 | NR_049788.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148429.hg.1 | TC0200007064.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200148428.hg.1 | ENST00000233575.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200148428.hg.1 | ENST00000427123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200148428.hg.1 | ENST00000440760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200148428.hg.1 | ENST00000453453.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200148428.hg.1 | ENST00000464279.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200148428.hg.1 | ENST00000484886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200148428.hg.1 | ENST00000489402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200148428.hg.1 | ENST00000494893.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200148428.hg.1 | ENST00000537606.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200148428.hg.1 | NM_001267059.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148428.hg.1 | NM_001267060.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148428.hg.1 | NM_014748.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148428.hg.1 | NR_049782.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148428.hg.1 | NR_049783.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148428.hg.1 | NR_049784.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148428.hg.1 | NR_049785.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148428.hg.1 | NR_049786.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148428.hg.1 | NR_049787.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148428.hg.1 | NR_049788.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148428.hg.1 | ENST00000233575.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200148428.hg.1 | ENST00000427123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200148428.hg.1 | ENST00000440760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200148428.hg.1 | ENST00000453453.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200148428.hg.1 | ENST00000464279.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200148428.hg.1 | ENST00000484886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200148428.hg.1 | ENST00000489402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200148428.hg.1 | ENST00000494893.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200148428.hg.1 | ENST00000537606.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200148428.hg.1 | NM_001267059.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148428.hg.1 | NM_001267060.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148428.hg.1 | NM_014748.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148428.hg.1 | NR_049782.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148428.hg.1 | NR_049783.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148428.hg.1 | NR_049784.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148428.hg.1 | NR_049785.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148428.hg.1 | NR_049786.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148428.hg.1 | NR_049787.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148428.hg.1 | NR_049788.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148428.hg.1 | TC0200007064.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200148418.hg.1 | ENST00000233575.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200148418.hg.1 | ENST00000537606.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200148418.hg.1 | NM_001267059.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148418.hg.1 | NM_001267060.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148418.hg.1 | NM_001267061.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148418.hg.1 | NM_014748.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148418.hg.1 | NR_049782.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148418.hg.1 | NR_049783.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148418.hg.1 | NR_049784.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148418.hg.1 | NR_049785.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148418.hg.1 | NR_049786.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148418.hg.1 | NR_049787.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148418.hg.1 | NR_049788.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148418.hg.1 | ENST00000233575.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200148418.hg.1 | ENST00000537606.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200148418.hg.1 | NM_001267059.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148418.hg.1 | NM_001267060.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148418.hg.1 | NM_001267061.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148418.hg.1 | NM_014748.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148418.hg.1 | NR_049782.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148418.hg.1 | NR_049783.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148418.hg.1 | NR_049784.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148418.hg.1 | NR_049785.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148418.hg.1 | NR_049786.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148418.hg.1 | NR_049787.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148418.hg.1 | NR_049788.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148418.hg.1 | TC0200007064.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200148439.hg.1 | ENST00000233575.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200148439.hg.1 | ENST00000427123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200148439.hg.1 | ENST00000440760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200148439.hg.1 | ENST00000453453.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200148439.hg.1 | ENST00000464279.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200148439.hg.1 | ENST00000494893.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200148439.hg.1 | ENST00000537606.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200148439.hg.1 | NM_001267059.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148439.hg.1 | NM_001267060.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148439.hg.1 | NM_001267061.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148439.hg.1 | NM_014748.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148439.hg.1 | NR_049782.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148439.hg.1 | NR_049783.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148439.hg.1 | NR_049784.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148439.hg.1 | NR_049785.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148439.hg.1 | NR_049786.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148439.hg.1 | NR_049787.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148439.hg.1 | NR_049788.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148439.hg.1 | ENST00000233575.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200148439.hg.1 | ENST00000427123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200148439.hg.1 | ENST00000440760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200148439.hg.1 | ENST00000453453.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200148439.hg.1 | ENST00000464279.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200148439.hg.1 | ENST00000494893.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200148439.hg.1 | ENST00000537606.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200148439.hg.1 | NM_001267059.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148439.hg.1 | NM_001267060.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148439.hg.1 | NM_001267061.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148439.hg.1 | NM_014748.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148439.hg.1 | NR_049782.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148439.hg.1 | NR_049783.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148439.hg.1 | NR_049784.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148439.hg.1 | NR_049785.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148439.hg.1 | NR_049786.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148439.hg.1 | NR_049787.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148439.hg.1 | NR_049788.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148439.hg.1 | TC0200007064.hg.1 | Clariom D TC |

SNX17
Genomic Map