Assay Details
| Gene Symbol: LOC100130476 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_049793.1 | 2 - 3 | 893 | 108 | ||
| GenBank mRNA | AK124173.1 | - | 2 - 3 | 893 | 108 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC100507406 | LOC100507406 | -126.1kb | Sense | Chr. 6 - 137730170 - 137739037 on NCBI Build GRCh38 | |
| Downstream |
PERP, TP53 apoptosis effector |
PERP |
KCP1, KRTCAP1, PIGPC1, THW, dJ496H19.1 |
221.6kb | Sense | Chr. 6 - 138088505 - 138107523 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000431144.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000431144.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000431144.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000431144.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000440578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000440578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000440578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000448942.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000448942.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000448942.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000448942.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000448942.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000448942.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000448942.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000448942.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000448942.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000448942.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000448942.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000448942.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000448942.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000448942.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000448942.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000448942.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000606327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000606327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000606327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000606998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000606998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000606998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000606998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000606998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000606998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000607671.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000607671.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000607671.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000607671.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000607671.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000607671.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000607671.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | NR_049793.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | NR_049793.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | NR_049793.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | NR_049793.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | NR_049793.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | NR_049793.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600192157.hg.1 | ENST00000606998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600192157.hg.1 | NR_049793.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600192157.hg.1 | ENST00000606998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600192157.hg.1 | NR_049793.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600192157.hg.1 | TC0600013316.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000431144.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000431144.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000431144.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000431144.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000440578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000440578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000440578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000448942.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000448942.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000448942.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000448942.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000448942.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000448942.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000448942.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000448942.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000448942.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000448942.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000448942.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000448942.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000448942.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000448942.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000448942.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000448942.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000606327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000606327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000606327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000606998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000606998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000606998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000606998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000606998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000606998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000607671.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000607671.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000607671.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000607671.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000607671.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000607671.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000607671.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | NR_049793.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | NR_049793.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | NR_049793.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | NR_049793.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | NR_049793.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | NR_049793.1 | Refseq |

LOC100130476
Genomic Map