Assay Details
| Gene Symbol: MIR5000 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_049796.1 | 1 | 50 | 74 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
DNA polymerase epsilon 4, accessory subunit |
POLE4 |
YHHQ1, p12 |
-121.1kb | Sense | Chr. 2 - 74958648 - 74969732 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC105374811 | LOC105374811 | 55.4kb | Sense | Chr. 2 - 75146352 - 75279143 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC0200008110.hg.1
Set Membership: |
|

MIR5000
Genomic Map