Assay Details
| Gene Symbol: MIR5008 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_049806.1 | 1 | 45 | 74 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
protease, serine 38 |
PRSS38 | MPN2 | -81.2kb | Anti-sense | Chr. 1 - 227815708 - 227846470 on NCBI Build GRCh38 |
| Downstream |
nascent polypeptide-associated complex subunit alpha, muscle-specific form-like |
LOC101060022 | 28.1kb | Anti-sense | Chr. 1 - 227968754 - 227971943 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC0100017577.hg.1
Set Membership: |
|

MIR5008
Genomic Map